The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126340.2 | NP_001119812.1 | 3 - 4 | 480 | 111 |
NM_001271818.1 | NP_001258747.1 | 3 - 4 | 503 | 111 | |
NM_032831.3 | NP_116220.1 | 2 - 3 | 371 | 111 | |
GenBank mRNA | AK027639.1 | - | 2 - 3 | 325 | 111 |
AK056497.1 | - | 3 - 4 | 111 | ||
BC069270.1 | - | 2 - 3 | 274 | 111 | |
CN366874.1 | - | 3 - 4 | 399 | 111 | |
DA770788.1 | - | 3 - 4 | 476 | 111 |