The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253891.1 | NP_001240820.1 | 3 - 4 | 563 | 62 |
NM_032564.4 | NP_115953.2 | 4 - 5 | 692 | 62 | |
XM_011545304.2 | XP_011543606.1 | 4 - 5 | 396 | 62 | |
GenBank mRNA | AB048286.2 | - | 4 - 5 | 663 | 62 |
AB073384.1 | - | 3 - 4 | 240 | 62 | |
AF384161.2 | - | 4 - 5 | 440 | 62 | |
AL834287.1 | - | 4 - 5 | 571 | 62 | |
AY358532.1 | - | 4 - 5 | 617 | 62 | |
AY780647.1 | - | 3 - 4 | 295 | 62 | |
BC015234.1 | - | 4 - 5 | 681 | 62 |