Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75640520-75641025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKDD1B
Entrez Gene ID
728780
Gene Name
ankyrin repeat and death domain containing 1B
Gene Aliases
-
UniGene
Hs.370455
Gene Chromosome Location
Chr.5: 75611289-75671846 on Build GRCh38
Transcripts
NM_001276713.1 XM_011543618.2 XM_011543623.2 XM_017009814.1 XM_017009815.1 XM_017009816.1 XM_017009817.1

SNP Detail

SNP ID Chromosome Location
rs977678853 Chr.5: 75640809-75640809 on Build GRCh38
rs1024216660 Chr.5: 75640545-75640545 on Build GRCh38
rs972371378 Chr.5: 75640705-75640705 on Build GRCh38
rs150402840 Chr.5: 75640989-75640989 on Build GRCh38
rs1014192272 Chr.5: 75640680-75640680 on Build GRCh38
rs757596823 Chr.5: 75640604-75640604 on Build GRCh38
rs73764949 Chr.5: 75640785-75640785 on Build GRCh38
rs936166187 Chr.5: 75640874-75640874 on Build GRCh38
rs373914108 Chr.5: 75640609-75640609 on Build GRCh38
rs1016730376 Chr.5: 75640704-75640704 on Build GRCh38
rs1028486281 Chr.5: 75640730-75640730 on Build GRCh38
rs950152753 Chr.5: 75640975-75640975 on Build GRCh38
rs918370165 Chr.5: 75640710-75640710 on Build GRCh38
rs547089898 Chr.5: 75640727-75640727 on Build GRCh38
rs887115169 Chr.5: 75640990-75640990 on Build GRCh38
rs1056275250 Chr.5: 75640622-75640622 on Build GRCh38
rs901200328 Chr.5: 75640763-75640763 on Build GRCh38
rs180811996 Chr.5: 75640973-75640973 on Build GRCh38
rs745999957 Chr.5: 75640846-75640846 on Build GRCh38
rs996722992 Chr.5: 75640710-75640710 on Build GRCh38
rs926260202 Chr.5: 75640812-75640812 on Build GRCh38
rs140482806 Chr.5: 75640954-75640954 on Build GRCh38
rs539263888 Chr.5: 75640752-75640752 on Build GRCh38
rs1003813490 Chr.5: 75640974-75640974 on Build GRCh38
rs936066170 Chr.5: 75640596-75640596 on Build GRCh38
rs1045739923 Chr.5: 75640692-75640692 on Build GRCh38
rs563844388 Chr.5: 75640543-75640543 on Build GRCh38
rs935358066 Chr.5: 75640780-75640780 on Build GRCh38
rs1011406470 Chr.5: 75640857-75640857 on Build GRCh38
rs78018749 Chr.5: 75640628-75640628 on Build GRCh38
rs188157472 Chr.5: 75640615-75640615 on Build GRCh38
rs942628405 Chr.5: 75640741-75640741 on Build GRCh38
rs984009757 Chr.5: 75640735-75640735 on Build GRCh38
rs557980284 Chr.5: 75641000-75641000 on Build GRCh38
rs544658078 Chr.5: 75640761-75640761 on Build GRCh38
rs527427661 Chr.5: 75640642-75640642 on Build GRCh38
rs1023809955 Chr.5: 75640934-75640934 on Build GRCh38
rs931046794 Chr.5: 75640731-75640731 on Build GRCh38
rs77302640 Chr.5: 75640571-75640571 on Build GRCh38
rs982323538 Chr.5: 75640578-75640578 on Build GRCh38
rs562013067 Chr.5: 75640701-75640704 on Build GRCh38
rs913583678 Chr.5: 75640767-75640767 on Build GRCh38
rs1055992256 Chr.5: 75640903-75640903 on Build GRCh38
rs751797383 Chr.5: 75640713-75640713 on Build GRCh38
rs886998502 Chr.5: 75640696-75640696 on Build GRCh38
rs535723358 Chr.5: 75640801-75640801 on Build GRCh38
rs148328508 Chr.5: 75640739-75640739 on Build GRCh38

View Full Product Details