The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170697.1 | NP_001164168.1 | 1 - 2 | 110 | 109 |
GenBank mRNA | AF367472.1 | - | 1 - 2 | 58 | 109 |
BP371122.1 | - | 1 - 2 | 110 | 109 |