Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78038799-78039295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3B1
Entrez Gene ID
8546
Gene Name
adaptor related protein complex 3 beta 1 subunit
Gene Aliases
ADTB3, ADTB3A, HPS, HPS2, PE
UniGene
Hs.532091
Gene Chromosome Location
Chr.5: 78002326-78294755 on Build GRCh38
Transcripts
NM_001271769.1 NM_003664.4 XM_005248618.3 XM_017010001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256323 c.2734C>T p.R912* Chr.5: 78039118-78039118 on Build GRCh38
6435361 c.2759G>A p.G920E Chr.5: 78039093-78039093 on Build GRCh38
272344 c.2762A>G p.E921G Chr.5: 78039090-78039090 on Build GRCh38
5918576 c.2773C>T p.P925S Chr.5: 78039079-78039079 on Build GRCh38
7199774 c.2784G>A p.M928I Chr.5: 78039068-78039068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024013390 Chr.5: 78038835-78038835 on Build GRCh38
rs747843144 Chr.5: 78039039-78039039 on Build GRCh38
rs574048736 Chr.5: 78039088-78039088 on Build GRCh38
rs776019289 Chr.5: 78038935-78038935 on Build GRCh38
rs558325834 Chr.5: 78039076-78039076 on Build GRCh38
rs946033247 Chr.5: 78038940-78038940 on Build GRCh38
rs375205110 Chr.5: 78039200-78039200 on Build GRCh38
rs1003245963 Chr.5: 78039010-78039010 on Build GRCh38
rs752519227 Chr.5: 78039118-78039118 on Build GRCh38
rs773298750 Chr.5: 78039226-78039226 on Build GRCh38
rs746532723 Chr.5: 78039049-78039049 on Build GRCh38
rs755942138 Chr.5: 78039017-78039017 on Build GRCh38
rs578144417 Chr.5: 78039079-78039079 on Build GRCh38
rs869312835 Chr.5: 78039150-78039150 on Build GRCh38
rs753810749 Chr.5: 78039090-78039090 on Build GRCh38
rs866933996 Chr.5: 78039216-78039216 on Build GRCh38
rs139344924 Chr.5: 78039191-78039191 on Build GRCh38
rs753619790 Chr.5: 78039019-78039019 on Build GRCh38
rs866308304 Chr.5: 78039072-78039072 on Build GRCh38
rs780503803 Chr.5: 78039181-78039181 on Build GRCh38
rs771283896 Chr.5: 78039069-78039069 on Build GRCh38
rs781759536 Chr.5: 78039162-78039162 on Build GRCh38
rs756994406 Chr.5: 78039271-78039271 on Build GRCh38
rs928845608 Chr.5: 78038832-78038832 on Build GRCh38
rs750433644 Chr.5: 78039156-78039157 on Build GRCh38
rs770455413 Chr.5: 78039179-78039179 on Build GRCh38
rs200985176 Chr.5: 78039085-78039085 on Build GRCh38
rs951058281 Chr.5: 78039027-78039027 on Build GRCh38
rs774631707 Chr.5: 78039143-78039143 on Build GRCh38
rs999392510 Chr.5: 78038968-78038968 on Build GRCh38
rs932217351 Chr.5: 78038932-78038932 on Build GRCh38
rs746328055 Chr.5: 78039163-78039163 on Build GRCh38
rs902217735 Chr.5: 78038952-78038952 on Build GRCh38
rs878879619 Chr.5: 78038958-78038958 on Build GRCh38
rs770332717 Chr.5: 78039075-78039075 on Build GRCh38
rs565017353 Chr.5: 78038929-78038929 on Build GRCh38
rs1049922800 Chr.5: 78038925-78038925 on Build GRCh38
rs765162490 Chr.5: 78039238-78039238 on Build GRCh38
rs151295181 Chr.5: 78038871-78038871 on Build GRCh38
rs770247199 Chr.5: 78038877-78038877 on Build GRCh38
rs77541448 Chr.5: 78038826-78038826 on Build GRCh38
rs763066332 Chr.5: 78038992-78038992 on Build GRCh38
rs778503399 Chr.5: 78039035-78039035 on Build GRCh38
rs763647354 Chr.5: 78039256-78039256 on Build GRCh38
rs561977108 Chr.5: 78039132-78039132 on Build GRCh38
rs192217190 Chr.5: 78038997-78038997 on Build GRCh38
rs746205404 Chr.5: 78039073-78039073 on Build GRCh38
rs1056281695 Chr.5: 78039265-78039265 on Build GRCh38
rs774705050 Chr.5: 78039209-78039209 on Build GRCh38
rs777107402 Chr.5: 78039046-78039046 on Build GRCh38
rs765309616 Chr.5: 78039031-78039032 on Build GRCh38
rs570046898 Chr.5: 78038874-78038874 on Build GRCh38
rs893622555 Chr.5: 78038988-78038988 on Build GRCh38
rs754832294 Chr.5: 78039029-78039029 on Build GRCh38
rs764958808 Chr.5: 78039095-78039095 on Build GRCh38
rs1052438449 Chr.5: 78038934-78038934 on Build GRCh38
rs759300647 Chr.5: 78039094-78039094 on Build GRCh38
rs745403781 Chr.5: 78038867-78038867 on Build GRCh38
rs146624866 Chr.5: 78039267-78039267 on Build GRCh38
rs970174552 Chr.5: 78039002-78039002 on Build GRCh38
rs766356766 Chr.5: 78039018-78039018 on Build GRCh38
rs756834284 Chr.5: 78039152-78039152 on Build GRCh38
rs75230657 Chr.5: 78038923-78038923 on Build GRCh38
rs746192704 Chr.5: 78038850-78038850 on Build GRCh38
rs761878887 Chr.5: 78039184-78039184 on Build GRCh38
rs758088280 Chr.5: 78039045-78039045 on Build GRCh38
rs762861855 Chr.5: 78039241-78039241 on Build GRCh38
rs772186737 Chr.5: 78039225-78039225 on Build GRCh38
rs761764690 Chr.5: 78039212-78039212 on Build GRCh38
rs763355104 Chr.5: 78038976-78038976 on Build GRCh38
rs143527588 Chr.5: 78039122-78039122 on Build GRCh38
rs751695483 Chr.5: 78038998-78038998 on Build GRCh38
rs777332984 Chr.5: 78039070-78039070 on Build GRCh38
rs535786508 Chr.5: 78038920-78038920 on Build GRCh38
rs144420604 Chr.5: 78039239-78039239 on Build GRCh38
rs572558916 Chr.5: 78038983-78038983 on Build GRCh38
rs201179527 Chr.5: 78039083-78039083 on Build GRCh38

View Full Product Details