The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201484.1 | NP_001188413.1 | 8 - 9 | 1837 | 69 |
NM_152850.3 | NP_690577.2 | 7 - 8 | 1385 | 69 | |
XM_017015223.1 | XP_016870712.1 | 7 - 8 | 1766 | 69 | |
XM_017015224.1 | XP_016870713.1 | 7 - 8 | 2030 | 69 | |
GenBank mRNA | BC001030.2 | - | 4 - 5 | 715 | 69 |
BC013987.2 | - | 5 - 6 | 801 | 69 | |
BC036916.1 | - | 6 - 7 | 1350 | 69 | |
BC065282.1 | - | 8 - 9 | 1508 | 69 |