The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026761.2 | 1 - 2 | 171 | 93 | |
GenBank mRNA | AK130857.1 | - | 1 - 2 | 138 | 93 |
AK130858.1 | - | 1 - 2 | 123 | 93 | |
BC005997.1 | - | 1 - 2 | 107 | 93 | |
BP197632.1 | - | 1 - 2 | 171 | 93 | |
BX382054.2 | - | 1 - 2 | 106 | 93 |