Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPUSD4
Entrez Gene ID
84881
Gene Name
RNA pseudouridylate synthase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 126202094 - 126211692 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144827.1 NP_001138299.1 3 - 4 609 100
NM_032795.2 NP_116184.2 3 - 4 609 100
GenBank mRNA AK027400.1 - 3 - 4 563 100
AK225320.1 - 3 - 4 569 100
AK308059.1 - 3 - 4 562 100
BC014131.2 - 3 - 4 570 100

View Full Product Details