Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93618030-93618538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
UniGene
Hs.600086
Gene Chromosome Location
Chr.5: 93615130-94111699 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1

SNP Detail

SNP ID Chromosome Location
rs964351014 Chr.5: 93618254-93618254 on Build GRCh38
rs909591016 Chr.5: 93618365-93618365 on Build GRCh38
rs761216854 Chr.5: 93618447-93618447 on Build GRCh38
rs566120989 Chr.5: 93618276-93618276 on Build GRCh38
rs879381147 Chr.5: 93618140-93618140 on Build GRCh38
rs554638129 Chr.5: 93618166-93618166 on Build GRCh38
rs776002951 Chr.5: 93618258-93618258 on Build GRCh38
rs959444628 Chr.5: 93618327-93618327 on Build GRCh38
rs998469850 Chr.5: 93618112-93618112 on Build GRCh38
rs768296601 Chr.5: 93618066-93618067 on Build GRCh38
rs1022988534 Chr.5: 93618236-93618236 on Build GRCh38
rs570112434 Chr.5: 93618064-93618064 on Build GRCh38
rs1051322047 Chr.5: 93618488-93618488 on Build GRCh38
rs957483701 Chr.5: 93618230-93618230 on Build GRCh38
rs1003671273 Chr.5: 93618151-93618151 on Build GRCh38
rs3180425 Chr.5: 93618071-93618071 on Build GRCh38
rs574542141 Chr.5: 93618251-93618251 on Build GRCh38
rs890056114 Chr.5: 93618503-93618503 on Build GRCh38
rs1011136616 Chr.5: 93618205-93618205 on Build GRCh38
rs948066885 Chr.5: 93618104-93618104 on Build GRCh38
rs1010614149 Chr.5: 93618272-93618272 on Build GRCh38
rs1029820092 Chr.5: 93618138-93618138 on Build GRCh38
rs73135347 Chr.5: 93618199-93618199 on Build GRCh38
rs918283836 Chr.5: 93618417-93618417 on Build GRCh38
rs183840193 Chr.5: 93618323-93618323 on Build GRCh38
rs1801627 Chr.5: 93618184-93618184 on Build GRCh38
rs1033481450 Chr.5: 93618213-93618213 on Build GRCh38
rs920576020 Chr.5: 93618065-93618065 on Build GRCh38
rs897374931 Chr.5: 93618108-93618112 on Build GRCh38
rs982497417 Chr.5: 93618362-93618362 on Build GRCh38
rs187353632 Chr.5: 93618406-93618406 on Build GRCh38
rs775669428 Chr.5: 93618486-93618486 on Build GRCh38
rs191811088 Chr.5: 93618483-93618483 on Build GRCh38
rs193079272 Chr.5: 93618206-93618206 on Build GRCh38
rs764556129 Chr.5: 93618313-93618313 on Build GRCh38
rs772269377 Chr.5: 93618145-93618145 on Build GRCh38
rs879127160 Chr.5: 93618293-93618293 on Build GRCh38
rs1007434818 Chr.5: 93618513-93618513 on Build GRCh38
rs1038257963 Chr.5: 93618063-93618063 on Build GRCh38
rs765006087 Chr.5: 93618491-93618491 on Build GRCh38
rs941125651 Chr.5: 93618400-93618400 on Build GRCh38
rs1020281465 Chr.5: 93618274-93618274 on Build GRCh38
rs906596295 Chr.5: 93618150-93618150 on Build GRCh38
rs908349216 Chr.5: 93618389-93618389 on Build GRCh38
rs567778665 Chr.5: 93618463-93618463 on Build GRCh38
rs761132137 Chr.5: 93618289-93618289 on Build GRCh38
rs1052568534 Chr.5: 93618193-93618193 on Build GRCh38
rs536755362 Chr.5: 93618473-93618473 on Build GRCh38
rs1036708851 Chr.5: 93618073-93618073 on Build GRCh38

View Full Product Details