Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93620609-93621126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
UniGene
Hs.600086
Gene Chromosome Location
Chr.5: 93615130-94111699 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NR_031754.1

Gene Symbol
MIR2277
Entrez Gene ID
100313887
Gene Name
microRNA 2277
Gene Aliases
mir-2277
UniGene
Gene Chromosome Location
Chr.5: 93620696-93620788 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NR_031754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366202 c.1125G>A p.E375E Chr.5: 93621113-93621113 on Build GRCh38
6658923 c.1176C>G p.A392A Chr.5: 93621062-93621062 on Build GRCh38
4137885 c.1159A>T p.K387* Chr.5: 93621079-93621079 on Build GRCh38
1071248 c.1185C>G p.A395A Chr.5: 93621053-93621053 on Build GRCh38
1311396 c.1114G>A p.D372N Chr.5: 93621124-93621124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377016885 Chr.5: 93620824-93620824 on Build GRCh38
rs897244214 Chr.5: 93620735-93620735 on Build GRCh38
rs994970632 Chr.5: 93620713-93620713 on Build GRCh38
rs1042890783 Chr.5: 93620705-93620705 on Build GRCh38
rs748568837 Chr.5: 93621074-93621074 on Build GRCh38
rs779244112 Chr.5: 93621068-93621068 on Build GRCh38
rs542094067 Chr.5: 93620696-93620696 on Build GRCh38
rs573452491 Chr.5: 93620755-93620755 on Build GRCh38
rs763461366 Chr.5: 93620990-93620990 on Build GRCh38
rs1008172445 Chr.5: 93620722-93620722 on Build GRCh38
rs746445655 Chr.5: 93620972-93620972 on Build GRCh38
rs1027121461 Chr.5: 93620757-93620757 on Build GRCh38
rs975762093 Chr.5: 93620689-93620689 on Build GRCh38
rs948349186 Chr.5: 93620935-93620935 on Build GRCh38
rs147889267 Chr.5: 93620917-93620918 on Build GRCh38
rs760054457 Chr.5: 93620756-93620766 on Build GRCh38
rs756104458 Chr.5: 93621029-93621029 on Build GRCh38
rs3210586 Chr.5: 93620677-93620677 on Build GRCh38
rs773825296 Chr.5: 93620950-93620951 on Build GRCh38
rs966001124 Chr.5: 93620659-93620659 on Build GRCh38
rs994214609 Chr.5: 93620738-93620738 on Build GRCh38
rs759366475 Chr.5: 93620726-93620726 on Build GRCh38
rs751516972 Chr.5: 93621016-93621016 on Build GRCh38
rs752660442 Chr.5: 93620750-93620750 on Build GRCh38
rs978924520 Chr.5: 93620979-93620979 on Build GRCh38
rs770325506 Chr.5: 93621090-93621090 on Build GRCh38
rs557932062 Chr.5: 93620992-93620992 on Build GRCh38
rs569135154 Chr.5: 93620969-93620969 on Build GRCh38
rs950784892 Chr.5: 93620645-93620645 on Build GRCh38
rs201849845 Chr.5: 93620970-93620970 on Build GRCh38
rs770507583 Chr.5: 93620974-93620974 on Build GRCh38
rs539903978 Chr.5: 93621077-93621077 on Build GRCh38
rs1040984878 Chr.5: 93621081-93621081 on Build GRCh38
rs111409602 Chr.5: 93620703-93620703 on Build GRCh38
rs748622169 Chr.5: 93620968-93620968 on Build GRCh38
rs965039743 Chr.5: 93620747-93620747 on Build GRCh38
rs753547235 Chr.5: 93620723-93620723 on Build GRCh38
rs549015536 Chr.5: 93620904-93620904 on Build GRCh38
rs371008338 Chr.5: 93620953-93620953 on Build GRCh38
rs765098130 Chr.5: 93620997-93620997 on Build GRCh38
rs912972273 Chr.5: 93620700-93620700 on Build GRCh38
rs553942281 Chr.5: 93620684-93620684 on Build GRCh38
rs978564179 Chr.5: 93620779-93620779 on Build GRCh38
rs1057256363 Chr.5: 93620911-93620911 on Build GRCh38
rs747181259 Chr.5: 93620758-93620758 on Build GRCh38
rs895500786 Chr.5: 93620709-93620709 on Build GRCh38
rs776321949 Chr.5: 93621091-93621091 on Build GRCh38
rs1034995780 Chr.5: 93620898-93620898 on Build GRCh38
rs745515122 Chr.5: 93621092-93621092 on Build GRCh38
rs974775994 Chr.5: 93620754-93620754 on Build GRCh38
rs764188036 Chr.5: 93620755-93620756 on Build GRCh38
rs754034568 Chr.5: 93621032-93621032 on Build GRCh38
rs763882914 Chr.5: 93621014-93621014 on Build GRCh38
rs987460239 Chr.5: 93620676-93620676 on Build GRCh38
rs775497647 Chr.5: 93620998-93620998 on Build GRCh38
rs181040414 Chr.5: 93620714-93620714 on Build GRCh38
rs911804319 Chr.5: 93621041-93621041 on Build GRCh38
rs776376048 Chr.5: 93620980-93620980 on Build GRCh38
rs1023614995 Chr.5: 93620711-93620711 on Build GRCh38
rs947185498 Chr.5: 93620701-93620701 on Build GRCh38
rs769631676 Chr.5: 93621095-93621095 on Build GRCh38
rs921201119 Chr.5: 93620691-93620694 on Build GRCh38
rs566442711 Chr.5: 93620866-93620866 on Build GRCh38
rs992518111 Chr.5: 93620687-93620687 on Build GRCh38
rs1019609621 Chr.5: 93620724-93620724 on Build GRCh38
rs890397500 Chr.5: 93620720-93620720 on Build GRCh38
rs755232147 Chr.5: 93621040-93621040 on Build GRCh38
rs779295050 Chr.5: 93620967-93620967 on Build GRCh38
rs977045862 Chr.5: 93621071-93621071 on Build GRCh38
rs758312601 Chr.5: 93620799-93620799 on Build GRCh38
rs934399306 Chr.5: 93620901-93620901 on Build GRCh38
rs749540545 Chr.5: 93620945-93620945 on Build GRCh38
rs79900553 Chr.5: 93621027-93621027 on Build GRCh38
rs758668942 Chr.5: 93620681-93620681 on Build GRCh38
rs1018931795 Chr.5: 93620742-93620742 on Build GRCh38
rs902934366 Chr.5: 93620704-93620704 on Build GRCh38
rs960307480 Chr.5: 93620903-93620903 on Build GRCh38
rs775297527 Chr.5: 93621101-93621101 on Build GRCh38
rs762937369 Chr.5: 93621008-93621008 on Build GRCh38
rs762591780 Chr.5: 93621104-93621104 on Build GRCh38
rs899416807 Chr.5: 93620712-93620712 on Build GRCh38
rs201013888 Chr.5: 93621078-93621078 on Build GRCh38
rs944665634 Chr.5: 93621079-93621079 on Build GRCh38
rs528867982 Chr.5: 93620867-93620867 on Build GRCh38
rs1056708335 Chr.5: 93620708-93620708 on Build GRCh38
rs573188534 Chr.5: 93620648-93620648 on Build GRCh38
rs546411947 Chr.5: 93620737-93620737 on Build GRCh38
rs980770846 Chr.5: 93620657-93620657 on Build GRCh38
rs559965517 Chr.5: 93621031-93621031 on Build GRCh38
rs750528963 Chr.5: 93620822-93620822 on Build GRCh38
rs952955666 Chr.5: 93620627-93620627 on Build GRCh38
rs1026361995 Chr.5: 93620629-93620629 on Build GRCh38
rs550720421 Chr.5: 93620710-93620710 on Build GRCh38
rs73772915 Chr.5: 93620716-93620716 on Build GRCh38
rs113720721 Chr.5: 93620921-93620922 on Build GRCh38
rs767634426 Chr.5: 93620817-93620817 on Build GRCh38
rs912939487 Chr.5: 93620926-93620926 on Build GRCh38
rs113403066 Chr.5: 93620924-93620925 on Build GRCh38
rs531124435 Chr.5: 93620706-93620706 on Build GRCh38
rs780071244 Chr.5: 93620936-93620936 on Build GRCh38

View Full Product Details