Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11orf70
Entrez Gene ID
85016
Gene Name
chromosome 11 open reading frame 70
Gene Aliases
-
Chromosome Location
Chr.11: 102047409 - 102084560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032930.2 NP_116319.2 4 - 5 469 97
XM_005271713.3 XP_005271770.1 4 - 5 497 97
GenBank mRNA AK094851.1 - 4 - 5 449 97
BC006128.1 - 3 - 4 673 97

View Full Product Details