The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033058.2 | NP_149047.2 | 2 - 3 | 568 | 73 |
NM_184085.1 | NP_908973.1 | 2 - 3 | 568 | 73 | |
NM_184086.1 | NP_908974.1 | 2 - 3 | 568 | 73 | |
NM_184087.1 | NP_908975.1 | 2 - 3 | 568 | 73 | |
XM_005251316.4 | XP_005251373.1 | 4 - 5 | 1593 | 73 | |
XM_011517613.2 | XP_011515915.1 | 4 - 5 | 1593 | 73 | |
XM_011517614.2 | XP_011515916.1 | 4 - 5 | 1516 | 73 | |
XM_011517615.2 | XP_011515917.1 | 4 - 5 | 1593 | 73 | |
XM_011517616.2 | XP_011515918.1 | 4 - 5 | 1593 | 73 | |
XM_017013908.1 | XP_016869397.1 | 2 - 3 | 286 | 73 | |
GenBank mRNA | AJ277493.1 | - | 2 - 3 | 568 | 73 |
AJ291712.1 | - | 2 - 3 | 474 | 73 | |
AJ431704.1 | - | 2 - 3 | 568 | 73 | |
AK091310.1 | - | 2 - 3 | 718 | 73 | |
AK091728.1 | - | 2 - 3 | 388 | 73 | |
BC007750.2 | - | 2 - 3 | 474 | 73 | |
BT007212.1 | - | 2 - 3 | 294 | 73 |