Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115841655-115842157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S1
Entrez Gene ID
1176
Gene Name
adaptor related protein complex 3 sigma 1 subunit
Gene Aliases
CLAPS3, Sigma3A
UniGene
Hs.406191
Gene Chromosome Location
Chr.5: 115841606-115914081 on Build GRCh38
Transcripts
NM_001002924.2 NM_001284.3 NM_001318090.1 NM_001318091.1 NM_001318093.1 NM_001318094.1 XM_011543136.2 XM_017009024.1 XM_017009025.1 NM_001277783.1 NM_004707.3 NR_033362.1 NR_033363.1 NR_073603.1

Gene Symbol
ATG12
Entrez Gene ID
9140
Gene Name
autophagy related 12
Gene Aliases
APG12, APG12L, FBR93, HAPG12
UniGene
Hs.264482
Gene Chromosome Location
Chr.5: 115828196-115841851 on Build GRCh38
Transcripts
NM_001002924.2 NM_001284.3 NM_001318090.1 NM_001318091.1 NM_001318093.1 NM_001318094.1 XM_011543136.2 XM_017009024.1 XM_017009025.1 NM_001277783.1 NM_004707.3 NR_033362.1 NR_033363.1 NR_073603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7323065 c.42G>A p.P14P Chr.5: 115842079-115842079 on Build GRCh38
299059 c.30C>A p.N10K Chr.5: 115842067-115842067 on Build GRCh38
6043875 c.64C>A p.P22T Chr.5: 115842101-115842101 on Build GRCh38
4422834 c.69_69+1insA p.S24fs*2 Chr.5: 115842106-115842107 on Build GRCh38
393689 c.39G>T p.K13N Chr.5: 115842076-115842076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545094953 Chr.5: 115841825-115841825 on Build GRCh38
rs867216094 Chr.5: 115841801-115841801 on Build GRCh38
rs768202808 Chr.5: 115842114-115842114 on Build GRCh38
rs779757861 Chr.5: 115842044-115842044 on Build GRCh38
rs576794189 Chr.5: 115841868-115841868 on Build GRCh38
rs764030106 Chr.5: 115842026-115842026 on Build GRCh38
rs537779984 Chr.5: 115841762-115841762 on Build GRCh38
rs773033432 Chr.5: 115842004-115842004 on Build GRCh38
rs11555658 Chr.5: 115842008-115842008 on Build GRCh38
rs973456490 Chr.5: 115841748-115841748 on Build GRCh38
rs572687640 Chr.5: 115841971-115841971 on Build GRCh38
rs369238013 Chr.5: 115842036-115842036 on Build GRCh38
rs775570888 Chr.5: 115842023-115842023 on Build GRCh38
rs985878605 Chr.5: 115841684-115841684 on Build GRCh38
rs746203087 Chr.5: 115841728-115841728 on Build GRCh38
rs965595104 Chr.5: 115841919-115841919 on Build GRCh38
rs555230454 Chr.5: 115841816-115841817 on Build GRCh38
rs141651630 Chr.5: 115842102-115842102 on Build GRCh38
rs368475651 Chr.5: 115842124-115842124 on Build GRCh38
rs752136463 Chr.5: 115842028-115842028 on Build GRCh38
rs890000473 Chr.5: 115841909-115841909 on Build GRCh38
rs894035460 Chr.5: 115841715-115841715 on Build GRCh38
rs958700920 Chr.5: 115841892-115841892 on Build GRCh38
rs779837175 Chr.5: 115841738-115841738 on Build GRCh38
rs751951920 Chr.5: 115841720-115841720 on Build GRCh38
rs62371472 Chr.5: 115841924-115841924 on Build GRCh38
rs935223106 Chr.5: 115841996-115841996 on Build GRCh38
rs1027617506 Chr.5: 115841880-115841880 on Build GRCh38
rs924415593 Chr.5: 115841791-115841792 on Build GRCh38
rs1050864 Chr.5: 115842024-115842024 on Build GRCh38
rs573906104 Chr.5: 115841824-115841824 on Build GRCh38
rs770647592 Chr.5: 115842084-115842084 on Build GRCh38
rs983960867 Chr.5: 115841826-115841826 on Build GRCh38
rs760442918 Chr.5: 115842009-115842009 on Build GRCh38
rs991395091 Chr.5: 115841893-115841893 on Build GRCh38
rs750873511 Chr.5: 115842039-115842039 on Build GRCh38
rs984259635 Chr.5: 115841959-115841959 on Build GRCh38
rs965398495 Chr.5: 115841799-115841799 on Build GRCh38
rs969279634 Chr.5: 115841982-115841982 on Build GRCh38
rs369070309 Chr.5: 115841691-115841691 on Build GRCh38
rs935764951 Chr.5: 115841798-115841798 on Build GRCh38
rs916642995 Chr.5: 115841981-115841981 on Build GRCh38
rs952773074 Chr.5: 115841954-115841954 on Build GRCh38
rs761967631 Chr.5: 115842111-115842111 on Build GRCh38
rs774362428 Chr.5: 115841988-115841988 on Build GRCh38
rs1052030241 Chr.5: 115841897-115841897 on Build GRCh38
rs773771496 Chr.5: 115841992-115841992 on Build GRCh38
rs1007454767 Chr.5: 115841910-115841910 on Build GRCh38
rs373869709 Chr.5: 115841697-115841697 on Build GRCh38
rs958588105 Chr.5: 115841754-115841754 on Build GRCh38
rs370092678 Chr.5: 115842085-115842085 on Build GRCh38
rs766607848 Chr.5: 115842139-115842139 on Build GRCh38
rs978647352 Chr.5: 115841932-115841932 on Build GRCh38
rs917051773 Chr.5: 115841763-115841763 on Build GRCh38
rs949946449 Chr.5: 115841775-115841775 on Build GRCh38
rs571157630 Chr.5: 115841778-115841778 on Build GRCh38
rs765415757 Chr.5: 115841700-115841700 on Build GRCh38
rs1046346204 Chr.5: 115841857-115841857 on Build GRCh38
rs74961482 Chr.5: 115841742-115841742 on Build GRCh38
rs534021522 Chr.5: 115841786-115841786 on Build GRCh38
rs771888426 Chr.5: 115841995-115841995 on Build GRCh38
rs747639202 Chr.5: 115841808-115841808 on Build GRCh38
rs200347946 Chr.5: 115842071-115842071 on Build GRCh38
rs755877400 Chr.5: 115841729-115841729 on Build GRCh38
rs377617825 Chr.5: 115842106-115842107 on Build GRCh38
rs748837438 Chr.5: 115842048-115842048 on Build GRCh38
rs999141079 Chr.5: 115841759-115841759 on Build GRCh38
rs772551972 Chr.5: 115841721-115841729 on Build GRCh38
rs11555657 Chr.5: 115841994-115841994 on Build GRCh38
rs781654617 Chr.5: 115841727-115841727 on Build GRCh38
rs769252301 Chr.5: 115841817-115841817 on Build GRCh38
rs776598809 Chr.5: 115841777-115841777 on Build GRCh38
rs539900902 Chr.5: 115841980-115841980 on Build GRCh38
rs1051830049 Chr.5: 115841868-115841878 on Build GRCh38
rs977255252 Chr.5: 115842086-115842086 on Build GRCh38
rs994940723 Chr.5: 115841878-115841878 on Build GRCh38
rs920607774 Chr.5: 115841749-115841749 on Build GRCh38
rs927923307 Chr.5: 115841989-115841989 on Build GRCh38
rs374408873 Chr.5: 115842096-115842096 on Build GRCh38
rs756538558 Chr.5: 115842042-115842042 on Build GRCh38
rs746872767 Chr.5: 115842077-115842077 on Build GRCh38
rs906431318 Chr.5: 115841884-115841884 on Build GRCh38
rs464020 Chr.5: 115841783-115841783 on Build GRCh38
rs796304723 Chr.5: 115841956-115841956 on Build GRCh38
rs932202879 Chr.5: 115841821-115841821 on Build GRCh38
rs370625498 Chr.5: 115842029-115842029 on Build GRCh38
rs1009390585 Chr.5: 115841730-115841730 on Build GRCh38
rs754543487 Chr.5: 115842049-115842049 on Build GRCh38
rs994181469 Chr.5: 115841920-115841920 on Build GRCh38
rs758395045 Chr.5: 115841705-115841705 on Build GRCh38
rs548944937 Chr.5: 115842135-115842135 on Build GRCh38
rs1022213811 Chr.5: 115841908-115841908 on Build GRCh38
rs186983208 Chr.5: 115841739-115841739 on Build GRCh38
rs1021767068 Chr.5: 115841780-115841780 on Build GRCh38
rs1020277421 Chr.5: 115841794-115841794 on Build GRCh38
rs1010577823 Chr.5: 115841905-115841905 on Build GRCh38
rs1019888433 Chr.5: 115841914-115841914 on Build GRCh38
rs144226661 Chr.5: 115841971-115841972 on Build GRCh38
rs78864570 Chr.5: 115841975-115841976 on Build GRCh38
rs761676325 Chr.5: 115841683-115841683 on Build GRCh38
rs954451332 Chr.5: 115841785-115841785 on Build GRCh38
rs144649545 Chr.5: 115841704-115841704 on Build GRCh38
rs944124891 Chr.5: 115841696-115841696 on Build GRCh38
rs554251951 Chr.5: 115841820-115841820 on Build GRCh38
rs149148380 Chr.5: 115841685-115841685 on Build GRCh38
rs778248571 Chr.5: 115842064-115842064 on Build GRCh38
rs189493290 Chr.5: 115841990-115841990 on Build GRCh38
rs757719374 Chr.5: 115841724-115841724 on Build GRCh38
rs774342712 Chr.5: 115842107-115842107 on Build GRCh38
rs774148498 Chr.5: 115841681-115841681 on Build GRCh38
rs765503917 Chr.5: 115842019-115842019 on Build GRCh38
rs916375901 Chr.5: 115841835-115841835 on Build GRCh38
rs374131819 Chr.5: 115842068-115842068 on Build GRCh38
rs780601857 Chr.5: 115841975-115841975 on Build GRCh38
rs556545516 Chr.5: 115841864-115841864 on Build GRCh38
rs569146156 Chr.5: 115841757-115841757 on Build GRCh38
rs935023684 Chr.5: 115841886-115841886 on Build GRCh38
rs573530420 Chr.5: 115841903-115841903 on Build GRCh38
rs565468163 Chr.5: 115841944-115841944 on Build GRCh38
rs1025613805 Chr.5: 115841921-115841921 on Build GRCh38
rs551141777 Chr.5: 115841776-115841776 on Build GRCh38
rs536673441 Chr.5: 115841851-115841851 on Build GRCh38
rs761131366 Chr.5: 115842132-115842132 on Build GRCh38
rs890617634 Chr.5: 115841833-115841833 on Build GRCh38
rs910354929 Chr.5: 115841915-115841915 on Build GRCh38
rs764729853 Chr.5: 115841709-115841709 on Build GRCh38
rs1048234620 Chr.5: 115841722-115841722 on Build GRCh38
rs935726762 Chr.5: 115842126-115842126 on Build GRCh38
rs768890009 Chr.5: 115842103-115842103 on Build GRCh38

View Full Product Details