The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171156.1 | NP_001164627.1 | 10 - 11 | 1874 | 76 |
NM_001171157.1 | NP_001164628.1 | 9 - 10 | 1763 | 76 | |
NM_001171158.1 | NP_001164629.1 | 9 - 10 | 1619 | 76 | |
NM_001171159.1 | NP_001164630.1 | 9 - 10 | 1589 | 76 | |
NM_001171161.1 | NP_001164632.1 | 8 - 9 | 1319 | 76 | |
NM_001322105.1 | NP_001309034.1 | 8 - 9 | 1493 | 76 | |
NM_033130.4 | NP_149121.2 | 10 - 11 | 2048 | 76 | |
XM_005259366.4 | XP_005259423.1 | 9 - 10 | 1606 | 76 | |
XM_005259367.3 | XP_005259424.1 | 9 - 10 | 1437 | 76 | |
XM_011527439.1 | XP_011525741.1 | 10 - 11 | 1732 | 76 | |
GenBank mRNA | AF301007.1 | - | 10 - 11 | 1943 | 76 |
AF310233.1 | - | 10 - 11 | 1863 | 76 | |
AF311905.1 | - | 9 - 10 | 1763 | 76 | |
AK289900.1 | - | 9 - 10 | 1549 | 76 | |
AK290542.1 | - | 9 - 10 | 1422 | 76 | |
AK292025.1 | - | 9 - 10 | 1666 | 76 | |
AK294185.1 | - | 9 - 10 | 1353 | 76 | |
AK303514.1 | - | 9 - 10 | 1458 | 76 | |
AK303717.1 | - | 8 - 9 | 1332 | 76 | |
AK308982.1 | - | 7 - 8 | 1429 | 76 | |
AY032685.1 | - | 10 - 11 | 1808 | 76 | |
AY358337.1 | - | 9 - 10 | 1412 | 76 | |
BC009955.2 | - | 6 - 7 | 818 | 76 | |
BC101725.1 | - | 10 - 11 | 1913 | 76 | |
BC143497.1 | - | 9 - 10 | 1628 | 76 | |
BC143500.1 | - | 10 - 11 | 1739 | 76 |