The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307941.1 | NP_001294870.1 | 1 - 2 | 179 | 85 |
NM_033280.3 | NP_150596.1 | 1 - 2 | 179 | 85 | |
GenBank mRNA | AF212233.1 | - | 1 - 2 | 144 | 85 |
AK295346.1 | - | 1 - 2 | 141 | 85 | |
AK307127.1 | - | 1 - 2 | 141 | 85 | |
AK314107.1 | - | 1 - 2 | 151 | 85 | |
BC009703.1 | - | 1 - 2 | 131 | 85 | |
BM789622.1 | - | 1 - 2 | 179 | 85 | |
BQ417397.1 | - | 1 - 2 | 72 | 85 |