Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131950807-131951349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL6
Entrez Gene ID
23305
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
ACS2, FACL6, LACS 6, LACS2, LACS5
UniGene
Hs.14945
Gene Chromosome Location
Chr.5: 131949973-132012068 on Build GRCh38
Transcripts
NM_001009185.2 NM_001205247.1 NM_001205248.1 NM_001205250.1 NM_001205251.1 NM_015256.3

SNP Detail

SNP ID Chromosome Location
rs928608266 Chr.5: 131951167-131951167 on Build GRCh38
rs79867633 Chr.5: 131951180-131951180 on Build GRCh38
rs903356929 Chr.5: 131951009-131951009 on Build GRCh38
rs938360928 Chr.5: 131950920-131950920 on Build GRCh38
rs921400039 Chr.5: 131951210-131951210 on Build GRCh38
rs947093723 Chr.5: 131951245-131951245 on Build GRCh38
rs648304 Chr.5: 131951015-131951015 on Build GRCh38
rs1007539379 Chr.5: 131951050-131951050 on Build GRCh38
rs914069330 Chr.5: 131950888-131950888 on Build GRCh38
rs988420007 Chr.5: 131951133-131951133 on Build GRCh38
rs367989916 Chr.5: 131951069-131951069 on Build GRCh38
rs909900688 Chr.5: 131951281-131951281 on Build GRCh38
rs750717886 Chr.5: 131950913-131950913 on Build GRCh38
rs17132127 Chr.5: 131951232-131951232 on Build GRCh38
rs903344495 Chr.5: 131951295-131951295 on Build GRCh38
rs186303217 Chr.5: 131950935-131950935 on Build GRCh38
rs558524487 Chr.5: 131950891-131950891 on Build GRCh38
rs537935663 Chr.5: 131950986-131950986 on Build GRCh38
rs567984083 Chr.5: 131951236-131951236 on Build GRCh38
rs999481243 Chr.5: 131951010-131951010 on Build GRCh38
rs1032028705 Chr.5: 131951057-131951057 on Build GRCh38
rs929859798 Chr.5: 131950899-131950899 on Build GRCh38
rs1030618195 Chr.5: 131951014-131951014 on Build GRCh38
rs555337890 Chr.5: 131950985-131950985 on Build GRCh38
rs573493181 Chr.5: 131951132-131951132 on Build GRCh38
rs940013875 Chr.5: 131951315-131951315 on Build GRCh38
rs1013415503 Chr.5: 131951026-131951026 on Build GRCh38
rs1042823076 Chr.5: 131951263-131951263 on Build GRCh38
rs1046976367 Chr.5: 131950901-131950901 on Build GRCh38
rs1025177045 Chr.5: 131951042-131951042 on Build GRCh38
rs553394655 Chr.5: 131951141-131951141 on Build GRCh38
rs1055495816 Chr.5: 131951217-131951217 on Build GRCh38
rs1017631611 Chr.5: 131951052-131951052 on Build GRCh38
rs904947170 Chr.5: 131951044-131951044 on Build GRCh38
rs983096120 Chr.5: 131951148-131951148 on Build GRCh38
rs867609679 Chr.5: 131950932-131950932 on Build GRCh38
rs894252581 Chr.5: 131950938-131950938 on Build GRCh38
rs919783546 Chr.5: 131950837-131950837 on Build GRCh38
rs919714459 Chr.5: 131951125-131951125 on Build GRCh38
rs192024577 Chr.5: 131950996-131950996 on Build GRCh38
rs530852216 Chr.5: 131951318-131951318 on Build GRCh38
rs921356550 Chr.5: 131950900-131950900 on Build GRCh38
rs973477459 Chr.5: 131951089-131951089 on Build GRCh38
rs542835909 Chr.5: 131951053-131951053 on Build GRCh38
rs771495354 Chr.5: 131950925-131950925 on Build GRCh38
rs944205994 Chr.5: 131950890-131950890 on Build GRCh38

View Full Product Details