Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131951993-131952504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL6
Entrez Gene ID
23305
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
ACS2, FACL6, LACS 6, LACS2, LACS5
UniGene
Hs.14945
Gene Chromosome Location
Chr.5: 131949973-132012068 on Build GRCh38
Transcripts
NM_001009185.2 NM_001205247.1 NM_001205248.1 NM_001205250.1 NM_001205251.1 NM_015256.3

SNP Detail

SNP ID Chromosome Location
rs915038560 Chr.5: 131952423-131952423 on Build GRCh38
rs982073634 Chr.5: 131952371-131952371 on Build GRCh38
rs973203275 Chr.5: 131952155-131952155 on Build GRCh38
rs955906017 Chr.5: 131952226-131952226 on Build GRCh38
rs191879490 Chr.5: 131952377-131952377 on Build GRCh38
rs540777245 Chr.5: 131952390-131952390 on Build GRCh38
rs560281984 Chr.5: 131952396-131952396 on Build GRCh38
rs1031296656 Chr.5: 131952067-131952067 on Build GRCh38
rs911681511 Chr.5: 131952247-131952247 on Build GRCh38
rs904359099 Chr.5: 131952481-131952481 on Build GRCh38
rs920772322 Chr.5: 131952333-131952333 on Build GRCh38
rs930885654 Chr.5: 131952336-131952336 on Build GRCh38
rs1053626737 Chr.5: 131952381-131952381 on Build GRCh38
rs943521234 Chr.5: 131952325-131952325 on Build GRCh38
rs895263279 Chr.5: 131952429-131952429 on Build GRCh38
rs886406368 Chr.5: 131952363-131952363 on Build GRCh38
rs1048448317 Chr.5: 131952353-131952353 on Build GRCh38
rs35992136 Chr.5: 131952095-131952096 on Build GRCh38
rs187177513 Chr.5: 131952227-131952227 on Build GRCh38
rs900867753 Chr.5: 131952445-131952445 on Build GRCh38
rs182907228 Chr.5: 131952409-131952409 on Build GRCh38
rs926523857 Chr.5: 131952372-131952372 on Build GRCh38
rs756344424 Chr.5: 131952432-131952432 on Build GRCh38
rs752121388 Chr.5: 131952279-131952279 on Build GRCh38
rs1056546744 Chr.5: 131952393-131952393 on Build GRCh38
rs974910712 Chr.5: 131952328-131952328 on Build GRCh38
rs555706416 Chr.5: 131952436-131952436 on Build GRCh38
rs188918735 Chr.5: 131952420-131952420 on Build GRCh38
rs979106069 Chr.5: 131952038-131952038 on Build GRCh38

View Full Product Details