Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131953167-131953716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL6
Entrez Gene ID
23305
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
ACS2, FACL6, LACS 6, LACS2, LACS5
UniGene
Hs.14945
Gene Chromosome Location
Chr.5: 131949973-132012068 on Build GRCh38
Transcripts
NM_001009185.2 NM_001205247.1 NM_001205248.1 NM_001205250.1 NM_001205251.1 NM_015256.3

SNP Detail

SNP ID Chromosome Location
rs893219336 Chr.5: 131953607-131953607 on Build GRCh38
rs1029920621 Chr.5: 131953435-131953436 on Build GRCh38
rs1040275017 Chr.5: 131953270-131953270 on Build GRCh38
rs973717680 Chr.5: 131953503-131953503 on Build GRCh38
rs566615199 Chr.5: 131953355-131953355 on Build GRCh38
rs897392222 Chr.5: 131953215-131953216 on Build GRCh38
rs151118803 Chr.5: 131953295-131953295 on Build GRCh38
rs368716175 Chr.5: 131953502-131953502 on Build GRCh38
rs887498067 Chr.5: 131953349-131953349 on Build GRCh38
rs944575660 Chr.5: 131953572-131953572 on Build GRCh38
rs531507915 Chr.5: 131953384-131953384 on Build GRCh38
rs893166995 Chr.5: 131953344-131953344 on Build GRCh38
rs776468960 Chr.5: 131953639-131953639 on Build GRCh38
rs1055929411 Chr.5: 131953595-131953595 on Build GRCh38
rs955220597 Chr.5: 131953464-131953464 on Build GRCh38
rs960872530 Chr.5: 131953373-131953373 on Build GRCh38
rs900457776 Chr.5: 131953675-131953675 on Build GRCh38
rs763812340 Chr.5: 131953410-131953410 on Build GRCh38
rs995823920 Chr.5: 131953285-131953285 on Build GRCh38
rs769683450 Chr.5: 131953283-131953283 on Build GRCh38
rs930239450 Chr.5: 131953543-131953543 on Build GRCh38
rs561337738 Chr.5: 131953426-131953426 on Build GRCh38
rs979869857 Chr.5: 131953451-131953451 on Build GRCh38
rs768294267 Chr.5: 131953568-131953568 on Build GRCh38
rs745849028 Chr.5: 131953221-131953221 on Build GRCh38
rs988244177 Chr.5: 131953506-131953506 on Build GRCh38
rs374070362 Chr.5: 131953485-131953485 on Build GRCh38
rs967039929 Chr.5: 131953405-131953405 on Build GRCh38
rs528146400 Chr.5: 131953523-131953523 on Build GRCh38
rs562763408 Chr.5: 131953356-131953356 on Build GRCh38
rs548128233 Chr.5: 131953385-131953385 on Build GRCh38
rs189104743 Chr.5: 131953558-131953558 on Build GRCh38
rs1015468109 Chr.5: 131953357-131953357 on Build GRCh38
rs1034421778 Chr.5: 131953317-131953317 on Build GRCh38
rs1011635528 Chr.5: 131953648-131953648 on Build GRCh38
rs925321087 Chr.5: 131953474-131953474 on Build GRCh38
rs944585345 Chr.5: 131953232-131953232 on Build GRCh38
rs935388932 Chr.5: 131953496-131953496 on Build GRCh38
rs985198395 Chr.5: 131953476-131953476 on Build GRCh38
rs903153666 Chr.5: 131953686-131953686 on Build GRCh38
rs1040263765 Chr.5: 131953588-131953588 on Build GRCh38
rs928825893 Chr.5: 131953569-131953569 on Build GRCh38
rs907346066 Chr.5: 131953294-131953294 on Build GRCh38
rs930208886 Chr.5: 131953236-131953236 on Build GRCh38

View Full Product Details