Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132659510-132660002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL13
Entrez Gene ID
3596
Gene Name
interleukin 13
Gene Aliases
IL-13, P600
UniGene
Hs.845
Gene Chromosome Location
Chr.5: 132658173-132661109 on Build GRCh38
Transcripts
NM_002188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317128 c.285G>A p.K95K Chr.5: 132659780-132659780 on Build GRCh38
373199 c.248C>A p.S83Y Chr.5: 132659743-132659743 on Build GRCh38
4692152 c.259G>A p.V87M Chr.5: 132659754-132659754 on Build GRCh38
5867259 c.331G>T p.G111W Chr.5: 132659826-132659826 on Build GRCh38
3137473 c.315G>A p.P105P Chr.5: 132659810-132659810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780855186 Chr.5: 132659783-132659783 on Build GRCh38
rs754520718 Chr.5: 132659837-132659846 on Build GRCh38
rs150226731 Chr.5: 132659735-132659735 on Build GRCh38
rs754514806 Chr.5: 132659680-132659680 on Build GRCh38
rs756707941 Chr.5: 132659744-132659744 on Build GRCh38
rs1030999463 Chr.5: 132659960-132659960 on Build GRCh38
rs773454019 Chr.5: 132659829-132659829 on Build GRCh38
rs1013002320 Chr.5: 132659531-132659531 on Build GRCh38
rs777431829 Chr.5: 132659767-132659767 on Build GRCh38
rs751035688 Chr.5: 132659829-132659830 on Build GRCh38
rs575861246 Chr.5: 132659905-132659905 on Build GRCh38
rs559233831 Chr.5: 132659879-132659879 on Build GRCh38
rs769087732 Chr.5: 132659798-132659798 on Build GRCh38
rs771386711 Chr.5: 132659688-132659688 on Build GRCh38
rs878869942 Chr.5: 132659584-132659584 on Build GRCh38
rs781754061 Chr.5: 132659823-132659824 on Build GRCh38
rs199513811 Chr.5: 132659755-132659755 on Build GRCh38
rs1035512579 Chr.5: 132659821-132659821 on Build GRCh38
rs538110009 Chr.5: 132659706-132659706 on Build GRCh38
rs200113399 Chr.5: 132659810-132659810 on Build GRCh38
rs532344450 Chr.5: 132659592-132659592 on Build GRCh38
rs776876174 Chr.5: 132659692-132659692 on Build GRCh38
rs747799114 Chr.5: 132659685-132659685 on Build GRCh38
rs2069747 Chr.5: 132659914-132659914 on Build GRCh38
rs201878455 Chr.5: 132659705-132659705 on Build GRCh38
rs952481319 Chr.5: 132659961-132659961 on Build GRCh38
rs878856925 Chr.5: 132659588-132659588 on Build GRCh38
rs752168427 Chr.5: 132659869-132659869 on Build GRCh38
rs73261671 Chr.5: 132659718-132659718 on Build GRCh38
rs544862168 Chr.5: 132659941-132659941 on Build GRCh38
rs746245700 Chr.5: 132659769-132659769 on Build GRCh38
rs1023014298 Chr.5: 132659591-132659591 on Build GRCh38
rs778748365 Chr.5: 132659681-132659681 on Build GRCh38
rs780429020 Chr.5: 132659636-132659636 on Build GRCh38
rs749795771 Chr.5: 132659792-132659792 on Build GRCh38
rs760436499 Chr.5: 132659848-132659848 on Build GRCh38
rs192837804 Chr.5: 132659729-132659729 on Build GRCh38
rs924151885 Chr.5: 132659618-132659618 on Build GRCh38
rs111692618 Chr.5: 132659970-132659970 on Build GRCh38
rs770108365 Chr.5: 132659774-132659774 on Build GRCh38
rs200545256 Chr.5: 132659834-132659834 on Build GRCh38
rs1028048466 Chr.5: 132659920-132659920 on Build GRCh38
rs778560262 Chr.5: 132659754-132659754 on Build GRCh38
rs544345482 Chr.5: 132659901-132659901 on Build GRCh38
rs201716976 Chr.5: 132659753-132659753 on Build GRCh38
rs757742023 Chr.5: 132659687-132659687 on Build GRCh38
rs1019451492 Chr.5: 132659896-132659896 on Build GRCh38
rs979393682 Chr.5: 132659918-132659919 on Build GRCh38
rs759548576 Chr.5: 132659858-132659858 on Build GRCh38
rs746179303 Chr.5: 132659700-132659700 on Build GRCh38
rs201744269 Chr.5: 132659860-132659860 on Build GRCh38
rs561270872 Chr.5: 132659969-132659969 on Build GRCh38
rs774429439 Chr.5: 132659799-132659799 on Build GRCh38
rs956632373 Chr.5: 132659635-132659635 on Build GRCh38
rs112070175 Chr.5: 132659849-132659849 on Build GRCh38
rs753892265 Chr.5: 132659855-132659855 on Build GRCh38
rs34654684 Chr.5: 132659731-132659731 on Build GRCh38
rs960863781 Chr.5: 132659583-132659583 on Build GRCh38
rs748829533 Chr.5: 132659676-132659676 on Build GRCh38
rs1000169601 Chr.5: 132659612-132659612 on Build GRCh38
rs868851412 Chr.5: 132659770-132659770 on Build GRCh38
rs892413770 Chr.5: 132659875-132659875 on Build GRCh38
rs200793410 Chr.5: 132659793-132659793 on Build GRCh38
rs780099128 Chr.5: 132659673-132659673 on Build GRCh38
rs553444004 Chr.5: 132659734-132659734 on Build GRCh38
rs764149458 Chr.5: 132659711-132659711 on Build GRCh38
rs200212730 Chr.5: 132659714-132659714 on Build GRCh38
rs552473475 Chr.5: 132659613-132659613 on Build GRCh38
rs201469726 Chr.5: 132659702-132659702 on Build GRCh38
rs1044510910 Chr.5: 132659833-132659833 on Build GRCh38
rs201804555 Chr.5: 132659809-132659809 on Build GRCh38
rs1009486089 Chr.5: 132659889-132659889 on Build GRCh38
rs374668631 Chr.5: 132659782-132659782 on Build GRCh38
rs200913948 Chr.5: 132659747-132659747 on Build GRCh38

View Full Product Details