Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132875727-132876238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF4
Entrez Gene ID
27125
Gene Name
AF4/FMR2 family member 4
Gene Aliases
AF5Q31, CHOPS, MCEF
UniGene
Hs.519313
Gene Chromosome Location
Chr.5: 132875379-132963662 on Build GRCh38
Transcripts
NM_014423.3 XM_005271963.4 XM_005271964.4 XM_006714587.3

SNP Detail

SNP ID Chromosome Location
rs988561370 Chr.5: 132876037-132876037 on Build GRCh38
rs960076097 Chr.5: 132875916-132875916 on Build GRCh38
rs1026603047 Chr.5: 132875827-132875827 on Build GRCh38
rs758270606 Chr.5: 132875884-132875884 on Build GRCh38
rs946932297 Chr.5: 132875871-132875871 on Build GRCh38
rs17166305 Chr.5: 132876163-132876163 on Build GRCh38
rs752143774 Chr.5: 132875993-132875993 on Build GRCh38
rs900066189 Chr.5: 132875777-132875777 on Build GRCh38
rs991319473 Chr.5: 132875931-132875931 on Build GRCh38
rs995745320 Chr.5: 132875796-132875796 on Build GRCh38
rs537386380 Chr.5: 132876106-132876106 on Build GRCh38
rs147288072 Chr.5: 132875950-132875950 on Build GRCh38
rs1035414441 Chr.5: 132875900-132875900 on Build GRCh38
rs1035315093 Chr.5: 132876155-132876157 on Build GRCh38
rs924237434 Chr.5: 132875926-132875926 on Build GRCh38
rs980066040 Chr.5: 132875872-132875872 on Build GRCh38
rs755706173 Chr.5: 132876097-132876097 on Build GRCh38
rs1039861676 Chr.5: 132876098-132876098 on Build GRCh38
rs912626125 Chr.5: 132876087-132876087 on Build GRCh38
rs772002964 Chr.5: 132875858-132875858 on Build GRCh38
rs924526140 Chr.5: 132875974-132875974 on Build GRCh38
rs532159340 Chr.5: 132875792-132875792 on Build GRCh38
rs571919442 Chr.5: 132875954-132875954 on Build GRCh38
rs1047382452 Chr.5: 132876085-132876085 on Build GRCh38
rs958439213 Chr.5: 132875767-132875767 on Build GRCh38
rs916726949 Chr.5: 132875862-132875862 on Build GRCh38
rs866640008 Chr.5: 132875969-132875969 on Build GRCh38
rs756779927 Chr.5: 132876060-132876060 on Build GRCh38
rs754650344 Chr.5: 132875932-132875936 on Build GRCh38
rs887371034 Chr.5: 132876200-132876200 on Build GRCh38
rs1021340211 Chr.5: 132876204-132876204 on Build GRCh38
rs931565065 Chr.5: 132876152-132876152 on Build GRCh38
rs566099754 Chr.5: 132875981-132875984 on Build GRCh38
rs1051105245 Chr.5: 132875932-132875932 on Build GRCh38
rs944144833 Chr.5: 132876094-132876094 on Build GRCh38
rs991235851 Chr.5: 132875816-132875816 on Build GRCh38
rs968615093 Chr.5: 132875955-132875955 on Build GRCh38

View Full Product Details