Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132880581-132881087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFF4
Entrez Gene ID
27125
Gene Name
AF4/FMR2 family member 4
Gene Aliases
AF5Q31, CHOPS, MCEF
UniGene
Hs.519313
Gene Chromosome Location
Chr.5: 132875379-132963662 on Build GRCh38
Transcripts
NM_014423.3 XM_005271963.4 XM_005271964.4 XM_006714587.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1294160 c.3466C>T p.R1156C Chr.5: 132881085-132881085 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569370320 Chr.5: 132880806-132880806 on Build GRCh38
rs986207895 Chr.5: 132880819-132880819 on Build GRCh38
rs1054915332 Chr.5: 132880652-132880652 on Build GRCh38
rs949285193 Chr.5: 132881017-132881017 on Build GRCh38
rs143386016 Chr.5: 132880995-132880996 on Build GRCh38
rs1032521084 Chr.5: 132880694-132880694 on Build GRCh38
rs1017669243 Chr.5: 132880859-132880859 on Build GRCh38
rs546506262 Chr.5: 132880905-132880905 on Build GRCh38
rs752012748 Chr.5: 132880904-132880904 on Build GRCh38
rs998076694 Chr.5: 132880789-132880789 on Build GRCh38
rs371660889 Chr.5: 132881043-132881043 on Build GRCh38
rs759395221 Chr.5: 132881032-132881032 on Build GRCh38
rs571675769 Chr.5: 132880614-132880614 on Build GRCh38
rs967615815 Chr.5: 132880770-132880770 on Build GRCh38
rs893283463 Chr.5: 132880698-132880698 on Build GRCh38
rs963472895 Chr.5: 132880884-132880884 on Build GRCh38
rs554403189 Chr.5: 132880800-132880800 on Build GRCh38
rs199585016 Chr.5: 132880715-132880716 on Build GRCh38
rs375967092 Chr.5: 132881049-132881049 on Build GRCh38
rs577456956 Chr.5: 132880816-132880816 on Build GRCh38
rs774965156 Chr.5: 132880994-132880994 on Build GRCh38
rs763598579 Chr.5: 132881064-132881064 on Build GRCh38
rs1030289756 Chr.5: 132880808-132880808 on Build GRCh38
rs113481875 Chr.5: 132880635-132880635 on Build GRCh38
rs1026450104 Chr.5: 132880933-132880933 on Build GRCh38
rs770779042 Chr.5: 132881019-132881019 on Build GRCh38
rs941839153 Chr.5: 132880928-132880928 on Build GRCh38
rs767716843 Chr.5: 132880972-132880972 on Build GRCh38
rs937689010 Chr.5: 132881004-132881004 on Build GRCh38
rs946564621 Chr.5: 132880681-132880681 on Build GRCh38
rs202005193 Chr.5: 132881038-132881038 on Build GRCh38
rs1011756458 Chr.5: 132880711-132880711 on Build GRCh38
rs1020514486 Chr.5: 132880745-132880745 on Build GRCh38
rs902440135 Chr.5: 132880782-132880782 on Build GRCh38
rs890493639 Chr.5: 132880814-132880814 on Build GRCh38
rs184033059 Chr.5: 132880976-132880976 on Build GRCh38
rs893547073 Chr.5: 132880676-132880676 on Build GRCh38
rs990466012 Chr.5: 132881005-132881005 on Build GRCh38
rs928193714 Chr.5: 132880953-132880953 on Build GRCh38
rs982424936 Chr.5: 132880942-132880942 on Build GRCh38
rs760394538 Chr.5: 132881045-132881045 on Build GRCh38
rs190942600 Chr.5: 132880651-132880651 on Build GRCh38
rs772526794 Chr.5: 132880809-132880809 on Build GRCh38
rs756504730 Chr.5: 132881024-132881024 on Build GRCh38
rs765177837 Chr.5: 132881036-132881036 on Build GRCh38
rs368107532 Chr.5: 132881026-132881026 on Build GRCh38
rs139233793 Chr.5: 132880952-132880952 on Build GRCh38
rs750786793 Chr.5: 132881014-132881014 on Build GRCh38
rs1042198989 Chr.5: 132880748-132880748 on Build GRCh38
rs762338359 Chr.5: 132880940-132880940 on Build GRCh38
rs937657919 Chr.5: 132880640-132880640 on Build GRCh38
rs781770655 Chr.5: 132880929-132880929 on Build GRCh38

View Full Product Details