The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000567.2 | NP_000558.2 | 1 - 2 | 162 | 72 |
XM_011509207.2 | XP_011507509.1 | 1 - 2 | 196 | 72 | |
GenBank mRNA | AK289443.1 | - | 1 - 2 | 162 | 72 |
AK313624.1 | - | 1 - 2 | 162 | 72 | |
BC020766.1 | - | 1 - 2 | 162 | 72 | |
BC125135.1 | - | 1 - 2 | 90 | 72 | |
BG533281.1 | - | - | 163 | 72 | |
X56214.1 | - | 1 - 2 | 147 | 72 | |
X56692.1 | - | 1 - 2 | 147 | 72 |