The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001999.3 | NP_001990.2 | 20 - 21 | 3115 | 74 |
XM_017009228.1 | XP_016864717.1 | 19 - 20 | 3278 | 74 | |
GenBank mRNA | AK300440.1 | - | 19 - 20 | 2795 | 74 |
U03272.1 | - | 20 - 21 | 2673 | 74 | |
X62009.1 | - | 4 - 5 | 420 | 74 |