The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002053.2 | NP_002044.2 | 1 - 2 | 200 | 72 |
GenBank mRNA | AB208912.1 | - | 1 - 2 | 159 | 72 |
AK291783.1 | - | 1 - 2 | 48 | 72 | |
AK298032.1 | - | 1 - 2 | 79 | 72 | |
BC002666.2 | - | 1 - 2 | 200 | 72 | |
M55542.1 | - | 1 - 2 | 48 | 72 |