The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098272.2 | NP_001091742.1 | 8 - 9 | 1407 | 120 |
NM_001324219.1 | NP_001311148.1 | 7 - 8 | 1394 | 120 | |
NM_001324220.1 | NP_001311149.1 | 8 - 9 | 1427 | 120 | |
NM_001324222.1 | NP_001311151.1 | 7 - 8 | 1281 | 120 | |
NM_001324223.1 | NP_001311152.1 | 6 - 7 | 1222 | 120 | |
NM_001324224.1 | NP_001311153.1 | 7 - 8 | 1301 | 120 | |
NM_002130.7 | NP_002121.4 | 7 - 8 | 1348 | 120 | |
XM_011514036.2 | XP_011512338.1 | 5 - 6 | 1213 | 120 | |
XM_017009412.1 | XP_016864901.1 | 8 - 9 | 1487 | 120 | |
GenBank mRNA | AK095492.1 | - | 8 - 9 | 1215 | 120 |
AK315593.1 | - | 8 - 9 | 1335 | 120 | |
BC000297.2 | - | 8 - 9 | 1286 | 120 | |
BC083514.1 | - | 7 - 8 | 1246 | 120 | |
BT007302.1 | - | 6 - 7 | 1181 | 120 | |
L25798.1 | - | 6 - 7 | 1202 | 120 | |
X66435.1 | - | 8 - 9 | 1303 | 120 |