Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148827690-148828196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRB2
Entrez Gene ID
154
Gene Name
adrenoceptor beta 2
Gene Aliases
ADRB2R, ADRBR, B2AR, BAR, BETA2AR
UniGene
Hs.2551
Gene Chromosome Location
Chr.5: 148826593-148828634 on Build GRCh38
Transcripts
NM_000024.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3211264 c.1138G>C p.D380H Chr.5: 148827969-148827969 on Build GRCh38
3211263 c.1028G>A p.R343H Chr.5: 148827859-148827859 on Build GRCh38
6350756 c.1239G>A p.L413L Chr.5: 148828070-148828070 on Build GRCh38
6303516 c.980G>T p.C327F Chr.5: 148827811-148827811 on Build GRCh38
3393327 c.1145C>T p.P382L Chr.5: 148827976-148827976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571344572 Chr.5: 148828117-148828117 on Build GRCh38
rs746492442 Chr.5: 148827832-148827832 on Build GRCh38
rs771585355 Chr.5: 148827955-148827955 on Build GRCh38
rs752216112 Chr.5: 148828014-148828014 on Build GRCh38
rs779607399 Chr.5: 148827830-148827830 on Build GRCh38
rs377115303 Chr.5: 148827741-148827741 on Build GRCh38
rs141761380 Chr.5: 148828104-148828105 on Build GRCh38
rs34522894 Chr.5: 148828097-148828097 on Build GRCh38
rs758318143 Chr.5: 148827827-148827827 on Build GRCh38
rs1042720 Chr.5: 148828070-148828070 on Build GRCh38
rs750591246 Chr.5: 148827767-148827767 on Build GRCh38
rs759505133 Chr.5: 148828096-148828096 on Build GRCh38
rs780905859 Chr.5: 148827936-148827936 on Build GRCh38
rs45580732 Chr.5: 148828099-148828100 on Build GRCh38
rs750235093 Chr.5: 148827996-148827996 on Build GRCh38
rs763433317 Chr.5: 148827869-148827869 on Build GRCh38
rs750228391 Chr.5: 148827922-148827922 on Build GRCh38
rs749645411 Chr.5: 148827953-148827953 on Build GRCh38
rs375373945 Chr.5: 148828096-148828096 on Build GRCh38
rs3730182 Chr.5: 148828010-148828010 on Build GRCh38
rs796421355 Chr.5: 148828097-148828098 on Build GRCh38
rs768294364 Chr.5: 148827733-148827733 on Build GRCh38
rs1026144198 Chr.5: 148828102-148828102 on Build GRCh38
rs143487925 Chr.5: 148827890-148827890 on Build GRCh38
rs766959341 Chr.5: 148827876-148827876 on Build GRCh38
rs374211656 Chr.5: 148828005-148828005 on Build GRCh38
rs751680275 Chr.5: 148827801-148827801 on Build GRCh38
rs45497201 Chr.5: 148828106-148828106 on Build GRCh38
rs372621803 Chr.5: 148828097-148828097 on Build GRCh38
rs41354346 Chr.5: 148827929-148827929 on Build GRCh38
rs145335962 Chr.5: 148827983-148827983 on Build GRCh38
rs1042719 Chr.5: 148827884-148827884 on Build GRCh38
rs41379548 Chr.5: 148828108-148828108 on Build GRCh38
rs370788074 Chr.5: 148828004-148828004 on Build GRCh38
rs779575155 Chr.5: 148828051-148828051 on Build GRCh38
rs762065041 Chr.5: 148827749-148827749 on Build GRCh38
rs752795911 Chr.5: 148827901-148827901 on Build GRCh38
rs149939087 Chr.5: 148827814-148827814 on Build GRCh38
rs767881740 Chr.5: 148827893-148827893 on Build GRCh38
rs756837704 Chr.5: 148828100-148828101 on Build GRCh38
rs780629446 Chr.5: 148828011-148828011 on Build GRCh38
rs796960157 Chr.5: 148828106-148828107 on Build GRCh38
rs145066318 Chr.5: 148827821-148827821 on Build GRCh38
rs41478145 Chr.5: 148828076-148828076 on Build GRCh38
rs746721501 Chr.5: 148827729-148827729 on Build GRCh38
rs780564175 Chr.5: 148828110-148828110 on Build GRCh38
rs866800666 Chr.5: 148827915-148827915 on Build GRCh38
rs201644897 Chr.5: 148827817-148827817 on Build GRCh38
rs377758965 Chr.5: 148827944-148827944 on Build GRCh38
rs376638463 Chr.5: 148828101-148828102 on Build GRCh38
rs766620962 Chr.5: 148827800-148827800 on Build GRCh38
rs993309698 Chr.5: 148827781-148827781 on Build GRCh38
rs963307282 Chr.5: 148827765-148827765 on Build GRCh38
rs748580918 Chr.5: 148827859-148827859 on Build GRCh38
rs755288362 Chr.5: 148827805-148827805 on Build GRCh38
rs989124620 Chr.5: 148827935-148827935 on Build GRCh38
rs146665903 Chr.5: 148827941-148827941 on Build GRCh38
rs764391729 Chr.5: 148828097-148828098 on Build GRCh38
rs4987254 Chr.5: 148828164-148828165 on Build GRCh38
rs746434318 Chr.5: 148828060-148828060 on Build GRCh38
rs369459230 Chr.5: 148827718-148827718 on Build GRCh38
rs879033030 Chr.5: 148827809-148827809 on Build GRCh38
rs752168351 Chr.5: 148828098-148828098 on Build GRCh38
rs751095383 Chr.5: 148827930-148827930 on Build GRCh38
rs777431854 Chr.5: 148828020-148828020 on Build GRCh38
rs914942592 Chr.5: 148828171-148828171 on Build GRCh38
rs767824140 Chr.5: 148827813-148827813 on Build GRCh38
rs771416666 Chr.5: 148827719-148827719 on Build GRCh38
rs748432014 Chr.5: 148828125-148828125 on Build GRCh38
rs373075228 Chr.5: 148827994-148827994 on Build GRCh38
rs776795902 Chr.5: 148828086-148828086 on Build GRCh38
rs41359451 Chr.5: 148828109-148828109 on Build GRCh38
rs774957439 Chr.5: 148827976-148827976 on Build GRCh38
rs542532864 Chr.5: 148828050-148828050 on Build GRCh38
rs774122860 Chr.5: 148828095-148828095 on Build GRCh38
rs371880712 Chr.5: 148827720-148827720 on Build GRCh38
rs779487214 Chr.5: 148828105-148828105 on Build GRCh38
rs767464563 Chr.5: 148828097-148828097 on Build GRCh38
rs369674098 Chr.5: 148827803-148827803 on Build GRCh38
rs6879202 Chr.5: 148828104-148828104 on Build GRCh38
rs768784387 Chr.5: 148828082-148828082 on Build GRCh38
rs758374900 Chr.5: 148827926-148827926 on Build GRCh38
rs140258602 Chr.5: 148827978-148827978 on Build GRCh38
rs776855632 Chr.5: 148828123-148828123 on Build GRCh38
rs768718219 Chr.5: 148828120-148828120 on Build GRCh38
rs200264766 Chr.5: 148828107-148828108 on Build GRCh38
rs989093317 Chr.5: 148828140-148828140 on Build GRCh38
rs766107767 Chr.5: 148828091-148828091 on Build GRCh38
rs764348560 Chr.5: 148827988-148827988 on Build GRCh38
rs756957418 Chr.5: 148828103-148828103 on Build GRCh38
rs755768214 Chr.5: 148828019-148828019 on Build GRCh38
rs750728100 Chr.5: 148828096-148828097 on Build GRCh38
rs761680537 Chr.5: 148827902-148827902 on Build GRCh38
rs1018502170 Chr.5: 148828142-148828142 on Build GRCh38
rs964337802 Chr.5: 148828152-148828152 on Build GRCh38
rs142595411 Chr.5: 148827790-148827790 on Build GRCh38
rs747091990 Chr.5: 148828112-148828112 on Build GRCh38
rs977299071 Chr.5: 148828166-148828166 on Build GRCh38
rs201717628 Chr.5: 148828107-148828107 on Build GRCh38
rs754447595 Chr.5: 148827932-148827932 on Build GRCh38
rs753482065 Chr.5: 148828100-148828100 on Build GRCh38
rs745657115 Chr.5: 148828103-148828104 on Build GRCh38
rs768349387 Chr.5: 148827850-148827850 on Build GRCh38
rs765797611 Chr.5: 148827750-148827750 on Build GRCh38
rs1012329753 Chr.5: 148827762-148827762 on Build GRCh38
rs28763957 Chr.5: 148828099-148828099 on Build GRCh38
rs773506135 Chr.5: 148828106-148828107 on Build GRCh38
rs368387516 Chr.5: 148828105-148828106 on Build GRCh38
rs377705165 Chr.5: 148828046-148828046 on Build GRCh38
rs201971211 Chr.5: 148828098-148828099 on Build GRCh38
rs3729944 Chr.5: 148828001-148828001 on Build GRCh38
rs753894727 Chr.5: 148827903-148827903 on Build GRCh38
rs773609492 Chr.5: 148827867-148827867 on Build GRCh38
rs145996440 Chr.5: 148827860-148827860 on Build GRCh38
rs374400618 Chr.5: 148827939-148827939 on Build GRCh38
rs749700176 Chr.5: 148828048-148828048 on Build GRCh38
rs762744199 Chr.5: 148828088-148828088 on Build GRCh38
rs759693360 Chr.5: 148827979-148827979 on Build GRCh38
rs776218939 Chr.5: 148827851-148827851 on Build GRCh38
rs111927761 Chr.5: 148828172-148828172 on Build GRCh38
rs772220983 Chr.5: 148827982-148827982 on Build GRCh38
rs577714514 Chr.5: 148827771-148827771 on Build GRCh38
rs1038401946 Chr.5: 148828079-148828079 on Build GRCh38
rs867664352 Chr.5: 148827912-148827912 on Build GRCh38

View Full Product Details