Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148989179-148989724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs1050578982 Chr.5: 148989387-148989387 on Build GRCh38
rs868605359 Chr.5: 148989542-148989542 on Build GRCh38
rs561106190 Chr.5: 148989613-148989613 on Build GRCh38
rs1038661336 Chr.5: 148989492-148989492 on Build GRCh38
rs946850230 Chr.5: 148989343-148989343 on Build GRCh38
rs540944260 Chr.5: 148989326-148989326 on Build GRCh38
rs573850536 Chr.5: 148989239-148989239 on Build GRCh38
rs940345017 Chr.5: 148989334-148989334 on Build GRCh38
rs987464500 Chr.5: 148989277-148989278 on Build GRCh38
rs530946516 Chr.5: 148989559-148989559 on Build GRCh38
rs189204132 Chr.5: 148989446-148989446 on Build GRCh38
rs769473499 Chr.5: 148989256-148989256 on Build GRCh38
rs776735616 Chr.5: 148989457-148989457 on Build GRCh38
rs886060115 Chr.5: 148989649-148989649 on Build GRCh38
rs903964728 Chr.5: 148989389-148989389 on Build GRCh38
rs1050613657 Chr.5: 148989633-148989633 on Build GRCh38
rs951883730 Chr.5: 148989555-148989555 on Build GRCh38
rs953431828 Chr.5: 148989579-148989579 on Build GRCh38
rs145210501 Chr.5: 148989476-148989476 on Build GRCh38
rs181229340 Chr.5: 148989586-148989586 on Build GRCh38
rs927080080 Chr.5: 148989296-148989296 on Build GRCh38
rs1040649701 Chr.5: 148989340-148989340 on Build GRCh38
rs979870879 Chr.5: 148989232-148989232 on Build GRCh38
rs975735570 Chr.5: 148989592-148989592 on Build GRCh38
rs770512169 Chr.5: 148989309-148989309 on Build GRCh38
rs957213011 Chr.5: 148989616-148989616 on Build GRCh38
rs563731773 Chr.5: 148989558-148989558 on Build GRCh38
rs115722370 Chr.5: 148989650-148989650 on Build GRCh38
rs147371811 Chr.5: 148989204-148989205 on Build GRCh38
rs1004129773 Chr.5: 148989479-148989479 on Build GRCh38
rs898334775 Chr.5: 148989525-148989525 on Build GRCh38
rs752686139 Chr.5: 148989258-148989258 on Build GRCh38
rs886060114 Chr.5: 148989400-148989400 on Build GRCh38
rs149989481 Chr.5: 148989224-148989224 on Build GRCh38
rs968203457 Chr.5: 148989680-148989680 on Build GRCh38
rs904034043 Chr.5: 148989649-148989649 on Build GRCh38
rs892482926 Chr.5: 148989423-148989423 on Build GRCh38
rs186004627 Chr.5: 148989328-148989328 on Build GRCh38
rs994931427 Chr.5: 148989229-148989229 on Build GRCh38
rs757547768 Chr.5: 148989509-148989509 on Build GRCh38
rs979945957 Chr.5: 148989500-148989500 on Build GRCh38
rs932201801 Chr.5: 148989621-148989621 on Build GRCh38
rs984500754 Chr.5: 148989316-148989316 on Build GRCh38
rs575869408 Chr.5: 148989662-148989662 on Build GRCh38
rs953377773 Chr.5: 148989246-148989246 on Build GRCh38
rs1031059021 Chr.5: 148989412-148989412 on Build GRCh38
rs971207143 Chr.5: 148989228-148989228 on Build GRCh38
rs939027930 Chr.5: 148989406-148989406 on Build GRCh38
rs746525833 Chr.5: 148989255-148989255 on Build GRCh38
rs1001346777 Chr.5: 148989394-148989394 on Build GRCh38
rs115194965 Chr.5: 148989222-148989222 on Build GRCh38
rs886060113 Chr.5: 148989372-148989372 on Build GRCh38
rs932170712 Chr.5: 148989372-148989372 on Build GRCh38
rs1016748267 Chr.5: 148989217-148989217 on Build GRCh38
rs915400636 Chr.5: 148989341-148989341 on Build GRCh38
rs902059813 Chr.5: 148989365-148989365 on Build GRCh38
rs1034132702 Chr.5: 148989515-148989515 on Build GRCh38
rs907578357 Chr.5: 148989572-148989572 on Build GRCh38
rs1029042941 Chr.5: 148989242-148989242 on Build GRCh38

View Full Product Details