Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148990446-148990989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs984555438 Chr.5: 148990964-148990964 on Build GRCh38
rs887712591 Chr.5: 148990512-148990512 on Build GRCh38
rs896718555 Chr.5: 148990639-148990639 on Build GRCh38
rs988700574 Chr.5: 148990901-148990901 on Build GRCh38
rs982237604 Chr.5: 148990813-148990813 on Build GRCh38
rs988428643 Chr.5: 148990762-148990762 on Build GRCh38
rs997313804 Chr.5: 148990557-148990557 on Build GRCh38
rs562719460 Chr.5: 148990628-148990628 on Build GRCh38
rs938076445 Chr.5: 148990849-148990849 on Build GRCh38
rs1030097117 Chr.5: 148990656-148990656 on Build GRCh38
rs530551992 Chr.5: 148990728-148990728 on Build GRCh38
rs912506879 Chr.5: 148990801-148990801 on Build GRCh38
rs191183936 Chr.5: 148990733-148990733 on Build GRCh38
rs548802448 Chr.5: 148990731-148990731 on Build GRCh38
rs943896722 Chr.5: 148990486-148990486 on Build GRCh38
rs1055395363 Chr.5: 148990582-148990582 on Build GRCh38
rs953100303 Chr.5: 148990658-148990658 on Build GRCh38
rs1053347453 Chr.5: 148990486-148990486 on Build GRCh38
rs532583447 Chr.5: 148990523-148990523 on Build GRCh38
rs914011706 Chr.5: 148990496-148990496 on Build GRCh38
rs374261707 Chr.5: 148990529-148990529 on Build GRCh38
rs912444172 Chr.5: 148990472-148990472 on Build GRCh38
rs947187227 Chr.5: 148990962-148990962 on Build GRCh38
rs571540820 Chr.5: 148990857-148990857 on Build GRCh38
rs761713767 Chr.5: 148990800-148990800 on Build GRCh38
rs534282709 Chr.5: 148990788-148990788 on Build GRCh38
rs1013778079 Chr.5: 148990695-148990695 on Build GRCh38
rs115296613 Chr.5: 148990513-148990513 on Build GRCh38
rs550732988 Chr.5: 148990623-148990623 on Build GRCh38
rs953709289 Chr.5: 148990764-148990764 on Build GRCh38
rs546889454 Chr.5: 148990850-148990850 on Build GRCh38
rs1031988612 Chr.5: 148990704-148990704 on Build GRCh38
rs926745562 Chr.5: 148990820-148990820 on Build GRCh38
rs1007678170 Chr.5: 148990681-148990681 on Build GRCh38
rs913048227 Chr.5: 148990945-148990945 on Build GRCh38
rs1026698259 Chr.5: 148990740-148990740 on Build GRCh38
rs764194691 Chr.5: 148990542-148990542 on Build GRCh38
rs751385381 Chr.5: 148990777-148990777 on Build GRCh38

View Full Product Details