Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149141400-149141900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM3
Entrez Gene ID
22885
Gene Name
actin binding LIM protein family member 3
Gene Aliases
HMFN1661
UniGene
Hs.49688
Gene Chromosome Location
Chr.5: 149141483-149260439 on Build GRCh38
Transcripts
NM_001301015.1 NM_001301027.1 NM_001301028.1 NM_014945.3 XM_005268392.1 XM_005268393.1 XM_005268394.1 XM_005268395.1 XM_005268396.1 XM_011537609.2 XM_017009238.1

SNP Detail

SNP ID Chromosome Location
rs979887636 Chr.5: 149141702-149141702 on Build GRCh38
rs1008501024 Chr.5: 149141789-149141789 on Build GRCh38
rs368707342 Chr.5: 149141484-149141484 on Build GRCh38
rs986725731 Chr.5: 149141616-149141616 on Build GRCh38
rs372347735 Chr.5: 149141755-149141755 on Build GRCh38
rs987122377 Chr.5: 149141422-149141422 on Build GRCh38
rs956121675 Chr.5: 149141813-149141813 on Build GRCh38
rs917184253 Chr.5: 149141865-149141865 on Build GRCh38
rs934495869 Chr.5: 149141842-149141842 on Build GRCh38
rs562762077 Chr.5: 149141500-149141500 on Build GRCh38
rs530295662 Chr.5: 149141530-149141530 on Build GRCh38
rs1002314425 Chr.5: 149141639-149141639 on Build GRCh38
rs1057152980 Chr.5: 149141523-149141523 on Build GRCh38
rs567309251 Chr.5: 149141614-149141614 on Build GRCh38
rs1032033782 Chr.5: 149141618-149141618 on Build GRCh38
rs955421654 Chr.5: 149141615-149141615 on Build GRCh38
rs940292923 Chr.5: 149141742-149141742 on Build GRCh38
rs914253624 Chr.5: 149141650-149141650 on Build GRCh38
rs763875976 Chr.5: 149141674-149141674 on Build GRCh38
rs1028396803 Chr.5: 149141780-149141780 on Build GRCh38
rs890947228 Chr.5: 149141782-149141782 on Build GRCh38
rs1038301819 Chr.5: 149141743-149141743 on Build GRCh38
rs774919236 Chr.5: 149141470-149141470 on Build GRCh38
rs914347035 Chr.5: 149141464-149141464 on Build GRCh38
rs1015850497 Chr.5: 149141689-149141689 on Build GRCh38
rs759935272 Chr.5: 149141622-149141622 on Build GRCh38
rs535119324 Chr.5: 149141626-149141626 on Build GRCh38
rs867953766 Chr.5: 149141546-149141546 on Build GRCh38
rs751549373 Chr.5: 149141599-149141599 on Build GRCh38
rs996897333 Chr.5: 149141596-149141596 on Build GRCh38
rs115496718 Chr.5: 149141772-149141772 on Build GRCh38
rs905368804 Chr.5: 149141513-149141513 on Build GRCh38
rs983062122 Chr.5: 149141458-149141458 on Build GRCh38
rs548610473 Chr.5: 149141737-149141737 on Build GRCh38
rs989790683 Chr.5: 149141623-149141623 on Build GRCh38
rs1015262394 Chr.5: 149141459-149141459 on Build GRCh38
rs955285999 Chr.5: 149141420-149141420 on Build GRCh38
rs887335505 Chr.5: 149141738-149141738 on Build GRCh38
rs190676273 Chr.5: 149141862-149141862 on Build GRCh38
rs926804890 Chr.5: 149141725-149141725 on Build GRCh38
rs1016909970 Chr.5: 149141551-149141551 on Build GRCh38
rs978514763 Chr.5: 149141803-149141803 on Build GRCh38
rs553021012 Chr.5: 149141669-149141669 on Build GRCh38
rs1028430401 Chr.5: 149141764-149141764 on Build GRCh38
rs905292543 Chr.5: 149141636-149141636 on Build GRCh38
rs952924897 Chr.5: 149141434-149141434 on Build GRCh38
rs1031520918 Chr.5: 149141791-149141791 on Build GRCh38
rs1004647068 Chr.5: 149141550-149141550 on Build GRCh38
rs978546201 Chr.5: 149141534-149141534 on Build GRCh38
rs1046887289 Chr.5: 149141633-149141633 on Build GRCh38
rs896781294 Chr.5: 149141437-149141460 on Build GRCh38
rs976869289 Chr.5: 149141497-149141497 on Build GRCh38
rs925786068 Chr.5: 149141824-149141824 on Build GRCh38
rs538631910 Chr.5: 149141846-149141846 on Build GRCh38
rs965112924 Chr.5: 149141494-149141494 on Build GRCh38
rs965383873 Chr.5: 149141730-149141730 on Build GRCh38
rs939617265 Chr.5: 149141517-149141517 on Build GRCh38
rs549153542 Chr.5: 149141553-149141553 on Build GRCh38
rs898507493 Chr.5: 149141749-149141749 on Build GRCh38
rs888653131 Chr.5: 149141673-149141673 on Build GRCh38

View Full Product Details