Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149237428-149237938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM3
Entrez Gene ID
22885
Gene Name
actin binding LIM protein family member 3
Gene Aliases
HMFN1661
UniGene
Hs.49688
Gene Chromosome Location
Chr.5: 149141483-149260439 on Build GRCh38
Transcripts
NM_001301015.1 NM_001301018.1 NM_001301027.1 NM_001301028.1 NM_014945.3 XM_005268392.1 XM_005268393.1 XM_005268394.1 XM_005268395.1 XM_005268396.1 XM_011537609.2 XM_017009238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4440026 c.1010C>A p.S337Y Chr.5: 149237569-149237569 on Build GRCh38
5474787 c.998C>A p.S333Y Chr.5: 149237557-149237557 on Build GRCh38
3852630 c.1027A>G p.R343G Chr.5: 149237586-149237586 on Build GRCh38
6031978 c.1044+6G>A p.? Chr.5: 149237609-149237609 on Build GRCh38
1064124 c.929C>T p.A310V Chr.5: 149237488-149237488 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543183223 Chr.5: 149237801-149237801 on Build GRCh38
rs1016218756 Chr.5: 149237694-149237694 on Build GRCh38
rs79537741 Chr.5: 149237696-149237696 on Build GRCh38
rs767959596 Chr.5: 149237553-149237553 on Build GRCh38
rs1046214817 Chr.5: 149237597-149237597 on Build GRCh38
rs772690429 Chr.5: 149237609-149237609 on Build GRCh38
rs34325217 Chr.5: 149237576-149237577 on Build GRCh38
rs766994899 Chr.5: 149237577-149237577 on Build GRCh38
rs764468102 Chr.5: 149237608-149237608 on Build GRCh38
rs772175512 Chr.5: 149237546-149237546 on Build GRCh38
rs575359991 Chr.5: 149237756-149237756 on Build GRCh38
rs757590344 Chr.5: 149237611-149237611 on Build GRCh38
rs779468104 Chr.5: 149237613-149237613 on Build GRCh38
rs545040628 Chr.5: 149237700-149237700 on Build GRCh38
rs1006340178 Chr.5: 149237900-149237900 on Build GRCh38
rs142756393 Chr.5: 149237869-149237869 on Build GRCh38
rs755753421 Chr.5: 149237604-149237604 on Build GRCh38
rs375261435 Chr.5: 149237760-149237761 on Build GRCh38
rs1037950039 Chr.5: 149237905-149237905 on Build GRCh38
rs919818624 Chr.5: 149237754-149237754 on Build GRCh38
rs981027803 Chr.5: 149237752-149237752 on Build GRCh38
rs1001100008 Chr.5: 149237855-149237855 on Build GRCh38
rs577594357 Chr.5: 149237874-149237874 on Build GRCh38
rs144195918 Chr.5: 149237573-149237573 on Build GRCh38
rs931244596 Chr.5: 149237760-149237760 on Build GRCh38
rs140543040 Chr.5: 149237570-149237570 on Build GRCh38
rs929686725 Chr.5: 149237870-149237870 on Build GRCh38
rs146590132 Chr.5: 149237601-149237601 on Build GRCh38
rs754479019 Chr.5: 149237507-149237507 on Build GRCh38
rs1013678026 Chr.5: 149237884-149237884 on Build GRCh38
rs766008911 Chr.5: 149237489-149237489 on Build GRCh38
rs765081609 Chr.5: 149237679-149237682 on Build GRCh38
rs758409346 Chr.5: 149237624-149237624 on Build GRCh38
rs923346520 Chr.5: 149237455-149237455 on Build GRCh38
rs780139117 Chr.5: 149237627-149237627 on Build GRCh38
rs756449768 Chr.5: 149237517-149237517 on Build GRCh38
rs556018790 Chr.5: 149237554-149237554 on Build GRCh38
rs749829462 Chr.5: 149237526-149237526 on Build GRCh38
rs150152582 Chr.5: 149237685-149237685 on Build GRCh38
rs886706088 Chr.5: 149237676-149237676 on Build GRCh38
rs989437626 Chr.5: 149237828-149237828 on Build GRCh38
rs898423039 Chr.5: 149237506-149237506 on Build GRCh38
rs376621261 Chr.5: 149237646-149237646 on Build GRCh38
rs779374635 Chr.5: 149237532-149237532 on Build GRCh38
rs149749871 Chr.5: 149237527-149237527 on Build GRCh38
rs916493872 Chr.5: 149237845-149237845 on Build GRCh38
rs765020392 Chr.5: 149237470-149237470 on Build GRCh38
rs557019341 Chr.5: 149237753-149237753 on Build GRCh38
rs977966888 Chr.5: 149237776-149237776 on Build GRCh38
rs762942384 Chr.5: 149237488-149237488 on Build GRCh38
rs750880702 Chr.5: 149237619-149237619 on Build GRCh38
rs563735987 Chr.5: 149237852-149237852 on Build GRCh38
rs202193990 Chr.5: 149237571-149237571 on Build GRCh38
rs940770892 Chr.5: 149237797-149237797 on Build GRCh38
rs761136342 Chr.5: 149237851-149237851 on Build GRCh38
rs751162589 Chr.5: 149237494-149237494 on Build GRCh38
rs947879230 Chr.5: 149237739-149237739 on Build GRCh38
rs145604284 Chr.5: 149237565-149237565 on Build GRCh38
rs759025870 Chr.5: 149237574-149237574 on Build GRCh38
rs773237860 Chr.5: 149237887-149237887 on Build GRCh38
rs79482203 Chr.5: 149237880-149237880 on Build GRCh38
rs537621133 Chr.5: 149237516-149237516 on Build GRCh38
rs947957052 Chr.5: 149237472-149237472 on Build GRCh38
rs745979758 Chr.5: 149237535-149237535 on Build GRCh38
rs201045172 Chr.5: 149237467-149237467 on Build GRCh38
rs775754253 Chr.5: 149237452-149237452 on Build GRCh38
rs529143894 Chr.5: 149237807-149237807 on Build GRCh38
rs535017363 Chr.5: 149237655-149237655 on Build GRCh38
rs540966774 Chr.5: 149237849-149237849 on Build GRCh38
rs954936763 Chr.5: 149237764-149237764 on Build GRCh38
rs752412260 Chr.5: 149237895-149237895 on Build GRCh38
rs954601039 Chr.5: 149237478-149237478 on Build GRCh38
rs569741861 Chr.5: 149237907-149237907 on Build GRCh38
rs745973261 Chr.5: 149237641-149237642 on Build GRCh38
rs781532306 Chr.5: 149237647-149237647 on Build GRCh38
rs201654077 Chr.5: 149237635-149237635 on Build GRCh38
rs980583248 Chr.5: 149237914-149237914 on Build GRCh38
rs762472416 Chr.5: 149237511-149237511 on Build GRCh38
rs901929884 Chr.5: 149237810-149237810 on Build GRCh38
rs1049079388 Chr.5: 149237781-149237781 on Build GRCh38
rs773283655 Chr.5: 149237479-149237479 on Build GRCh38
rs978312832 Chr.5: 149237483-149237483 on Build GRCh38
rs368112588 Chr.5: 149237551-149237551 on Build GRCh38
rs371689634 Chr.5: 149237566-149237566 on Build GRCh38
rs1030441502 Chr.5: 149237707-149237707 on Build GRCh38
rs886475751 Chr.5: 149237785-149237785 on Build GRCh38
rs996329650 Chr.5: 149237815-149237815 on Build GRCh38
rs559615001 Chr.5: 149237755-149237756 on Build GRCh38
rs138559616 Chr.5: 149237806-149237806 on Build GRCh38

View Full Product Details