The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322908.1 | NP_001309837.1 | 3 - 4 | 241 | |
NM_003081.4 | NP_003072.2 | 1 - 2 | 184 | 63 | |
NM_130811.3 | NP_570824.1 | 1 - 2 | 184 | 63 | |
XM_017028023.1 | XP_016883512.1 | 1 - 2 | 288 | 63 | |
GenBank mRNA | AK094560.1 | - | 1 - 2 | 152 | 63 |
AK098767.1 | - | 1 - 2 | 54 | 63 | |
AK223617.1 | - | 1 - 2 | 146 | 63 | |
AK314359.1 | - | 1 - 2 | 95 | 63 | |
BC010647.1 | - | 1 - 2 | 73 | 63 | |
BI552777.1 | - | 1 - 2 | 157 | 63 | |
BI602062.1 | - | 1 - 2 | 162 | 63 | |
BI604120.1 | - | 1 - 2 | 161 | 63 | |
BX647443.1 | - | 1 - 2 | 56 | 63 | |
D21267.1 | - | 1 - 2 | 146 | 63 | |
DA072367.1 | - | 1 - 2 | 184 | 63 | |
DA162693.1 | - | 1 - 2 | 288 | 63 |