Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157491516-157492013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM19
Entrez Gene ID
8728
Gene Name
ADAM metallopeptidase domain 19
Gene Aliases
FKSG34, MADDAM, MLTNB
UniGene
Hs.483944
Gene Chromosome Location
Chr.5: 157477304-157575823 on Build GRCh38
Transcripts
NM_033274.4 XM_005266003.2 XM_011534682.2 XM_017010009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770419 c.1954G>T p.E652* Chr.5: 157491873-157491873 on Build GRCh38
5770420 c.1147G>T p.E383* Chr.5: 157491873-157491873 on Build GRCh38
6227598 c.1108-7C>T p.? Chr.5: 157491919-157491919 on Build GRCh38
4659314 c.2065G>T p.G689W Chr.5: 157491651-157491651 on Build GRCh38
6170442 c.1932G>A p.R644R Chr.5: 157491889-157491889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755039969 Chr.5: 157491727-157491727 on Build GRCh38
rs569511696 Chr.5: 157491585-157491585 on Build GRCh38
rs771495678 Chr.5: 157491771-157491771 on Build GRCh38
rs764140640 Chr.5: 157491964-157491964 on Build GRCh38
rs372336872 Chr.5: 157491752-157491752 on Build GRCh38
rs780061330 Chr.5: 157491627-157491627 on Build GRCh38
rs11466790 Chr.5: 157491979-157491979 on Build GRCh38
rs753689875 Chr.5: 157491702-157491702 on Build GRCh38
rs941473999 Chr.5: 157491621-157491621 on Build GRCh38
rs763838612 Chr.5: 157491807-157491807 on Build GRCh38
rs10063366 Chr.5: 157491934-157491934 on Build GRCh38
rs762832265 Chr.5: 157491942-157491942 on Build GRCh38
rs749559925 Chr.5: 157491869-157491869 on Build GRCh38
rs746170380 Chr.5: 157491624-157491624 on Build GRCh38
rs372083563 Chr.5: 157491593-157491593 on Build GRCh38
rs762790571 Chr.5: 157491879-157491881 on Build GRCh38
rs749691225 Chr.5: 157491762-157491762 on Build GRCh38
rs555968256 Chr.5: 157491604-157491604 on Build GRCh38
rs374341398 Chr.5: 157491709-157491709 on Build GRCh38
rs777947434 Chr.5: 157491757-157491757 on Build GRCh38
rs140037727 Chr.5: 157491841-157491841 on Build GRCh38
rs778063164 Chr.5: 157491863-157491863 on Build GRCh38
rs146858323 Chr.5: 157491877-157491877 on Build GRCh38
rs182092067 Chr.5: 157491831-157491831 on Build GRCh38
rs535681267 Chr.5: 157491584-157491584 on Build GRCh38
rs760195529 Chr.5: 157491797-157491797 on Build GRCh38
rs752574745 Chr.5: 157491857-157491858 on Build GRCh38
rs773582493 Chr.5: 157491582-157491582 on Build GRCh38
rs952009424 Chr.5: 157491765-157491765 on Build GRCh38
rs763343405 Chr.5: 157491658-157491658 on Build GRCh38
rs771703929 Chr.5: 157491789-157491789 on Build GRCh38
rs904022524 Chr.5: 157491951-157491951 on Build GRCh38
rs758368854 Chr.5: 157491839-157491839 on Build GRCh38
rs190049336 Chr.5: 157491606-157491606 on Build GRCh38
rs1008257440 Chr.5: 157491965-157491965 on Build GRCh38
rs939579577 Chr.5: 157491640-157491640 on Build GRCh38
rs369351955 Chr.5: 157491919-157491919 on Build GRCh38
rs772728570 Chr.5: 157491918-157491918 on Build GRCh38
rs765072237 Chr.5: 157491834-157491834 on Build GRCh38
rs922400476 Chr.5: 157491540-157491540 on Build GRCh38
rs779487366 Chr.5: 157491898-157491898 on Build GRCh38
rs11466791 Chr.5: 157491780-157491780 on Build GRCh38
rs200883455 Chr.5: 157491832-157491832 on Build GRCh38
rs759120064 Chr.5: 157491679-157491679 on Build GRCh38
rs201100159 Chr.5: 157491768-157491768 on Build GRCh38
rs1012401723 Chr.5: 157491676-157491676 on Build GRCh38
rs139353150 Chr.5: 157491702-157491703 on Build GRCh38
rs773035417 Chr.5: 157491939-157491939 on Build GRCh38
rs752057338 Chr.5: 157491941-157491941 on Build GRCh38
rs139066384 Chr.5: 157491961-157491961 on Build GRCh38
rs748464152 Chr.5: 157491639-157491639 on Build GRCh38
rs761518159 Chr.5: 157491935-157491935 on Build GRCh38
rs775010615 Chr.5: 157491774-157491774 on Build GRCh38
rs375647154 Chr.5: 157491932-157491932 on Build GRCh38
rs746346343 Chr.5: 157491916-157491916 on Build GRCh38
rs2287749 Chr.5: 157491842-157491842 on Build GRCh38
rs767219173 Chr.5: 157491686-157491686 on Build GRCh38
rs150693665 Chr.5: 157491689-157491689 on Build GRCh38
rs921804703 Chr.5: 157491756-157491756 on Build GRCh38
rs762110868 Chr.5: 157491560-157491560 on Build GRCh38
rs1019568928 Chr.5: 157491694-157491694 on Build GRCh38
rs746434083 Chr.5: 157491785-157491785 on Build GRCh38
rs34882961 Chr.5: 157491630-157491630 on Build GRCh38
rs561948234 Chr.5: 157491838-157491838 on Build GRCh38
rs964047306 Chr.5: 157491725-157491725 on Build GRCh38
rs570671502 Chr.5: 157491631-157491631 on Build GRCh38
rs752847811 Chr.5: 157491854-157491854 on Build GRCh38
rs374726786 Chr.5: 157491796-157491796 on Build GRCh38
rs539487843 Chr.5: 157491552-157491552 on Build GRCh38
rs751160317 Chr.5: 157491587-157491587 on Build GRCh38
rs985018271 Chr.5: 157491776-157491776 on Build GRCh38
rs767307014 Chr.5: 157491603-157491603 on Build GRCh38
rs771583639 Chr.5: 157491659-157491659 on Build GRCh38
rs141977307 Chr.5: 157491652-157491652 on Build GRCh38
rs777681542 Chr.5: 157491609-157491609 on Build GRCh38
rs748291556 Chr.5: 157491739-157491739 on Build GRCh38
rs572323626 Chr.5: 157491623-157491623 on Build GRCh38
rs781408559 Chr.5: 157491637-157491637 on Build GRCh38
rs766375282 Chr.5: 157491947-157491947 on Build GRCh38
rs757103387 Chr.5: 157491704-157491704 on Build GRCh38
rs773956047 Chr.5: 157491660-157491660 on Build GRCh38
rs893393080 Chr.5: 157491651-157491651 on Build GRCh38
rs756253549 Chr.5: 157491861-157491861 on Build GRCh38
rs759213072 Chr.5: 157491600-157491600 on Build GRCh38
rs368768876 Chr.5: 157491735-157491735 on Build GRCh38
rs141502864 Chr.5: 157491779-157491779 on Build GRCh38
rs139027135 Chr.5: 157491688-157491688 on Build GRCh38
rs947690198 Chr.5: 157491920-157491920 on Build GRCh38
rs752656815 Chr.5: 157491960-157491960 on Build GRCh38
rs770280914 Chr.5: 157491567-157491567 on Build GRCh38
rs373569980 Chr.5: 157491929-157491929 on Build GRCh38
rs765166182 Chr.5: 157491708-157491708 on Build GRCh38

View Full Product Details