The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142548.1 | NP_001136020.1 | 9 - 10 | 1004 | 76 |
NM_003579.3 | NP_003570.2 | 8 - 9 | 1601 | 76 | |
XM_006710975.3 | XP_006711038.1 | 8 - 9 | 939 | 76 | |
XM_011542299.2 | XP_011540601.1 | 3 - 4 | 371 | 76 | |
XM_011542300.2 | XP_011540602.1 | 2 - 3 | 187 | 76 | |
GenBank mRNA | AK292611.1 | - | 9 - 10 | 984 | 76 |
AK297204.1 | - | 7 - 8 | 1088 | 76 | |
BC121059.1 | - | 8 - 9 | 1109 | 76 | |
BC121060.1 | - | 8 - 9 | 1109 | 76 | |
X97795.1 | - | 8 - 9 | 990 | 76 |