The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201563.5 | NP_963857.3 | 1 - 2 | 214 | 79 |
NR_047648.1 | 1 - 2 | 214 | 79 | ||
GenBank mRNA | AK308168.1 | - | 1 - 2 | 207 | 79 |
BC137397.1 | - | 1 - 2 | 157 | 79 | |
BC137423.1 | - | 1 - 2 | 157 | 79 | |
BP320976.1 | - | 1 - 2 | 205 | 79 | |
U90938.1 | - | 1 - 2 | 117 | 79 | |
U90939.1 | - | 1 - 2 | 117 | 79 | |
U90940.1 | - | 1 - 2 | 117 | 79 | |
U90941.1 | - | 1 - 2 | 117 | 79 | |
X17652.1 | - | 1 - 2 | 148 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002273.2 | NP_001002273.1 | 1 - 2 | 242 | 76 |
NM_001002274.2 | NP_001002274.1 | 1 - 2 | 242 | 79 | |
NM_001002275.2 | NP_001002275.1 | 1 - 2 | 242 | 76 | |
NM_004001.4 | NP_003992.3 | 1 - 2 | 242 | 79 | |
XM_011509292.2 | XP_011507594.1 | 2 - 3 | 206 | 79 | |
XM_017000669.1 | XP_016856158.1 | 2 - 3 | 206 | 79 | |
XM_017000670.1 | XP_016856159.1 | 1 - 2 | 187 | 76 | |
XM_017000671.1 | XP_016856160.1 | 2 - 3 | 206 | 79 | |
XM_017000672.1 | XP_016856161.1 | 2 - 3 | 206 | 79 | |
GenBank mRNA | AB209585.1 | - | 1 - 2 | 165 | 79 |
AF543826.1 | - | 1 - 2 | 195 | 79 | |
BC031992.1 | - | 1 - 2 | 152 | 79 | |
BC141826.1 | - | 1 - 2 | 152 | 79 | |
BC146678.1 | - | 1 - 2 | 157 | 79 | |
BC148273.1 | - | 1 - 2 | 137 | 79 | |
CB988406.1 | - | 1 - 2 | 234 | 79 | |
CR407635.1 | - | 1 - 2 | 115 | 79 | |
DC381078.1 | - | 1 - 2 | 242 | 79 | |
M28696.1 | - | 1 - 2 | 156 | 79 | |
M31934.1 | - | 1 - 2 | 154 | 79 | |
M31935.1 | - | 1 - 2 | 65 | 79 | |
U87560.1 | - | 1 - 2 | 115 | 79 | |
U87561.1 | - | 1 - 2 | 115 | 79 | |
U87562.1 | - | 1 - 2 | 115 | 79 | |
U87563.1 | - | 1 - 2 | 115 | 79 | |
U87564.1 | - | 1 - 2 | 115 | 79 | |
U87565.1 | - | 1 - 2 | 115 | 79 | |
U87566.1 | - | 1 - 2 | 115 | 79 | |
U87567.1 | - | 1 - 2 | 115 | 79 | |
U87568.1 | - | 1 - 2 | 115 | 79 | |
U87569.1 | - | 1 - 2 | 115 | 79 | |
U87570.1 | - | 1 - 2 | 115 | 76 | |
U87571.1 | - | 1 - 2 | 115 | 76 | |
U87572.1 | - | 1 - 2 | 115 | 76 | |
U87573.1 | - | 1 - 2 | 115 | 79 | |
U87574.1 | - | 1 - 2 | 115 | 79 | |
U87575.1 | - | 1 - 2 | 115 | 79 | |
U87576.1 | - | 1 - 2 | 115 | 79 | |
U87577.1 | - | 1 - 2 | 115 | 76 | |
X17653.1 | - | 1 - 2 | 184 | 79 | |
X52473.1 | - | 1 - 2 | 160 | 79 | |
X62573.1 | - | 1 - 2 | 79 |