Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 178222263-178222765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHYKPL
Entrez Gene ID
85007
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
Gene Aliases
AGXT2L2, PHLU
UniGene
Hs.248746
Gene Chromosome Location
Chr.5: 178208474-178232822 on Build GRCh38
Transcripts
NM_001278346.1 NM_153373.3 NR_103508.1 NR_103509.1 NR_103510.1 XM_006714925.2 XM_011534669.2 XM_011534670.2 XM_011534671.2 XM_011534672.2 XM_011534674.2 XM_011534676.2 XM_017009995.1 XM_017009996.1 XM_017009997.1 XM_017009998.1 XM_017009999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235645 c.910G>A p.V304I Chr.5: 178222372-178222372 on Build GRCh38
6988444 c.847G>A p.G283S Chr.5: 178222435-178222435 on Build GRCh38
3342014 c.846C>T p.N282N Chr.5: 178222436-178222436 on Build GRCh38
1244660 c.789G>C p.Q263H Chr.5: 178222493-178222493 on Build GRCh38
236967 c.909C>T p.G303G Chr.5: 178222373-178222373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543691949 Chr.5: 178222609-178222609 on Build GRCh38
rs760765398 Chr.5: 178222436-178222436 on Build GRCh38
rs78625692 Chr.5: 178222325-178222325 on Build GRCh38
rs746702673 Chr.5: 178222374-178222374 on Build GRCh38
rs1040281741 Chr.5: 178222378-178222378 on Build GRCh38
rs372439508 Chr.5: 178222419-178222419 on Build GRCh38
rs749623855 Chr.5: 178222388-178222388 on Build GRCh38
rs988848917 Chr.5: 178222724-178222724 on Build GRCh38
rs957374246 Chr.5: 178222483-178222483 on Build GRCh38
rs982163215 Chr.5: 178222626-178222626 on Build GRCh38
rs780887370 Chr.5: 178222544-178222544 on Build GRCh38
rs1023898998 Chr.5: 178222447-178222447 on Build GRCh38
rs749664390 Chr.5: 178222451-178222451 on Build GRCh38
rs780822431 Chr.5: 178222449-178222449 on Build GRCh38
rs776879679 Chr.5: 178222575-178222575 on Build GRCh38
rs910692662 Chr.5: 178222690-178222690 on Build GRCh38
rs752344333 Chr.5: 178222546-178222546 on Build GRCh38
rs762738037 Chr.5: 178222599-178222599 on Build GRCh38
rs772738952 Chr.5: 178222474-178222474 on Build GRCh38
rs375663899 Chr.5: 178222331-178222331 on Build GRCh38
rs774791401 Chr.5: 178222394-178222394 on Build GRCh38
rs971225845 Chr.5: 178222625-178222625 on Build GRCh38
rs779108954 Chr.5: 178222560-178222560 on Build GRCh38
rs555432504 Chr.5: 178222627-178222627 on Build GRCh38
rs1032986263 Chr.5: 178222684-178222684 on Build GRCh38
rs931909441 Chr.5: 178222604-178222604 on Build GRCh38
rs760851143 Chr.5: 178222496-178222496 on Build GRCh38
rs972116928 Chr.5: 178222653-178222653 on Build GRCh38
rs573701859 Chr.5: 178222651-178222651 on Build GRCh38
rs376845758 Chr.5: 178222617-178222617 on Build GRCh38
rs539750873 Chr.5: 178222509-178222509 on Build GRCh38
rs754564386 Chr.5: 178222446-178222446 on Build GRCh38
rs930860650 Chr.5: 178222663-178222663 on Build GRCh38
rs553923363 Chr.5: 178222344-178222344 on Build GRCh38
rs151042213 Chr.5: 178222412-178222412 on Build GRCh38
rs1036459922 Chr.5: 178222644-178222644 on Build GRCh38
rs779496779 Chr.5: 178222439-178222439 on Build GRCh38
rs201692484 Chr.5: 178222528-178222528 on Build GRCh38
rs766296013 Chr.5: 178222603-178222603 on Build GRCh38
rs141654643 Chr.5: 178222373-178222373 on Build GRCh38
rs367908733 Chr.5: 178222375-178222375 on Build GRCh38
rs568493755 Chr.5: 178222303-178222303 on Build GRCh38
rs141079773 Chr.5: 178222521-178222521 on Build GRCh38
rs768933736 Chr.5: 178222502-178222502 on Build GRCh38
rs758843042 Chr.5: 178222737-178222737 on Build GRCh38
rs368984594 Chr.5: 178222418-178222418 on Build GRCh38
rs977714710 Chr.5: 178222302-178222302 on Build GRCh38
rs375297594 Chr.5: 178222417-178222417 on Build GRCh38
rs753718086 Chr.5: 178222343-178222343 on Build GRCh38
rs755675967 Chr.5: 178222622-178222622 on Build GRCh38
rs896536001 Chr.5: 178222645-178222645 on Build GRCh38
rs751359897 Chr.5: 178222360-178222361 on Build GRCh38
rs1004956270 Chr.5: 178222428-178222428 on Build GRCh38
rs559399560 Chr.5: 178222682-178222682 on Build GRCh38
rs369759314 Chr.5: 178222333-178222333 on Build GRCh38
rs775258736 Chr.5: 178222293-178222293 on Build GRCh38
rs754554200 Chr.5: 178222533-178222533 on Build GRCh38
rs759651226 Chr.5: 178222395-178222395 on Build GRCh38
rs928086995 Chr.5: 178222643-178222643 on Build GRCh38
rs150267080 Chr.5: 178222565-178222565 on Build GRCh38
rs764023080 Chr.5: 178222335-178222335 on Build GRCh38
rs759543269 Chr.5: 178222327-178222327 on Build GRCh38
rs766388681 Chr.5: 178222462-178222462 on Build GRCh38
rs751089152 Chr.5: 178222443-178222443 on Build GRCh38
rs767510625 Chr.5: 178222589-178222591 on Build GRCh38
rs757045121 Chr.5: 178222587-178222588 on Build GRCh38
rs150108455 Chr.5: 178222696-178222696 on Build GRCh38
rs960880710 Chr.5: 178222442-178222442 on Build GRCh38
rs748277104 Chr.5: 178222587-178222587 on Build GRCh38
rs147023998 Chr.5: 178222404-178222404 on Build GRCh38
rs750375739 Chr.5: 178222435-178222435 on Build GRCh38
rs41285581 Chr.5: 178222396-178222396 on Build GRCh38
rs765616846 Chr.5: 178222572-178222572 on Build GRCh38
rs771261600 Chr.5: 178222455-178222455 on Build GRCh38
rs543595395 Chr.5: 178222294-178222294 on Build GRCh38
rs201105857 Chr.5: 178222564-178222564 on Build GRCh38
rs774160730 Chr.5: 178222376-178222376 on Build GRCh38
rs772822232 Chr.5: 178222591-178222591 on Build GRCh38
rs1013731059 Chr.5: 178222445-178222445 on Build GRCh38
rs373326864 Chr.5: 178222606-178222606 on Build GRCh38
rs751000339 Chr.5: 178222531-178222531 on Build GRCh38
rs763473941 Chr.5: 178222304-178222304 on Build GRCh38
rs56753982 Chr.5: 178222674-178222674 on Build GRCh38
rs920835282 Chr.5: 178222660-178222660 on Build GRCh38
rs771407068 Chr.5: 178222391-178222391 on Build GRCh38
rs372306666 Chr.5: 178222356-178222356 on Build GRCh38
rs1049557162 Chr.5: 178222621-178222621 on Build GRCh38
rs745378448 Chr.5: 178222370-178222370 on Build GRCh38
rs766813424 Chr.5: 178222316-178222316 on Build GRCh38
rs373675466 Chr.5: 178222522-178222522 on Build GRCh38
rs757902202 Chr.5: 178222372-178222372 on Build GRCh38
rs757708088 Chr.5: 178222610-178222611 on Build GRCh38
rs770074629 Chr.5: 178222590-178222590 on Build GRCh38
rs780710628 Chr.5: 178222566-178222566 on Build GRCh38
rs775826637 Chr.5: 178222408-178222408 on Build GRCh38
rs746404914 Chr.5: 178222465-178222465 on Build GRCh38
rs200141370 Chr.5: 178222573-178222573 on Build GRCh38
rs750347865 Chr.5: 178222592-178222593 on Build GRCh38
rs1001108480 Chr.5: 178222680-178222680 on Build GRCh38
rs765206289 Chr.5: 178222432-178222432 on Build GRCh38
rs774335119 Chr.5: 178222605-178222605 on Build GRCh38
rs775663443 Chr.5: 178222485-178222485 on Build GRCh38
rs1040083020 Chr.5: 178222726-178222726 on Build GRCh38

View Full Product Details