Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179614480-179614977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPH1
Entrez Gene ID
3187
Gene Name
heterogeneous nuclear ribonucleoprotein H1 (H)
Gene Aliases
HNRPH, HNRPH1, hnRNPH
UniGene
Hs.604001
Gene Chromosome Location
Chr.5: 179614178-179634784 on Build GRCh38
Transcripts
NM_001257293.1 NM_005520.2 XM_005265895.2 XM_005265896.2 XM_005265901.1 XM_005265902.4 XM_006714862.2 XM_006714863.2 XM_011534542.1 XM_011534544.1 XM_011534545.1 XM_011534546.1 XM_011534547.1 XM_017009414.1 XM_017009415.1 XM_017009416.1 XM_017009417.1 XM_017009418.1 XM_017009419.1 XM_017009420.1 XM_017009421.1 XM_017009422.1 XM_017009423.1 XM_017009424.1 XM_017009425.1

SNP Detail

SNP ID Chromosome Location
rs996027108 Chr.5: 179614743-179614743 on Build GRCh38
rs989874000 Chr.5: 179614788-179614788 on Build GRCh38
rs1057311102 Chr.5: 179614567-179614567 on Build GRCh38
rs948928012 Chr.5: 179614802-179614802 on Build GRCh38
rs895884219 Chr.5: 179614791-179614791 on Build GRCh38
rs908519165 Chr.5: 179614713-179614713 on Build GRCh38
rs989800653 Chr.5: 179614629-179614629 on Build GRCh38
rs1031337170 Chr.5: 179614613-179614617 on Build GRCh38
rs761797503 Chr.5: 179614807-179614814 on Build GRCh38
rs935783135 Chr.5: 179614601-179614601 on Build GRCh38
rs928542283 Chr.5: 179614689-179614689 on Build GRCh38
rs547250657 Chr.5: 179614670-179614670 on Build GRCh38
rs568496415 Chr.5: 179614752-179614760 on Build GRCh38
rs569243079 Chr.5: 179614811-179614811 on Build GRCh38
rs987592144 Chr.5: 179614628-179614629 on Build GRCh38
rs774838350 Chr.5: 179614594-179614594 on Build GRCh38
rs1008144408 Chr.5: 179614928-179614928 on Build GRCh38
rs774833761 Chr.5: 179614835-179614835 on Build GRCh38
rs955777263 Chr.5: 179614628-179614628 on Build GRCh38
rs369393320 Chr.5: 179614778-179614778 on Build GRCh38
rs539184494 Chr.5: 179614744-179614744 on Build GRCh38
rs1037314754 Chr.5: 179614728-179614728 on Build GRCh38
rs544306478 Chr.5: 179614915-179614915 on Build GRCh38
rs1004668469 Chr.5: 179614535-179614535 on Build GRCh38
rs576489734 Chr.5: 179614785-179614786 on Build GRCh38
rs978673993 Chr.5: 179614573-179614573 on Build GRCh38
rs767781648 Chr.5: 179614807-179614807 on Build GRCh38
rs199644228 Chr.5: 179614784-179614784 on Build GRCh38
rs968944625 Chr.5: 179614599-179614599 on Build GRCh38
rs376066558 Chr.5: 179614783-179614784 on Build GRCh38
rs558369023 Chr.5: 179614900-179614900 on Build GRCh38
rs10479547 Chr.5: 179614785-179614785 on Build GRCh38
rs536122560 Chr.5: 179614831-179614831 on Build GRCh38
rs902751315 Chr.5: 179614947-179614947 on Build GRCh38
rs1019843258 Chr.5: 179614529-179614529 on Build GRCh38
rs919779622 Chr.5: 179614864-179614864 on Build GRCh38
rs760180052 Chr.5: 179614837-179614837 on Build GRCh38
rs143667244 Chr.5: 179614588-179614588 on Build GRCh38
rs898574346 Chr.5: 179614741-179614741 on Build GRCh38
rs764170704 Chr.5: 179614878-179614878 on Build GRCh38
rs762154041 Chr.5: 179614901-179614901 on Build GRCh38
rs957124351 Chr.5: 179614787-179614787 on Build GRCh38
rs774551628 Chr.5: 179614803-179614804 on Build GRCh38
rs760776452 Chr.5: 179614877-179614877 on Build GRCh38
rs746739928 Chr.5: 179614857-179614857 on Build GRCh38
rs878955393 Chr.5: 179614784-179614785 on Build GRCh38
rs951973888 Chr.5: 179614942-179614942 on Build GRCh38
rs966875541 Chr.5: 179614572-179614572 on Build GRCh38
rs1032952363 Chr.5: 179614786-179614786 on Build GRCh38
rs869175916 Chr.5: 179614785-179614791 on Build GRCh38
rs777744279 Chr.5: 179614702-179614702 on Build GRCh38
rs891426571 Chr.5: 179614848-179614848 on Build GRCh38
rs1008196937 Chr.5: 179614762-179614762 on Build GRCh38
rs796072865 Chr.5: 179614805-179614806 on Build GRCh38
rs73336147 Chr.5: 179614906-179614906 on Build GRCh38
rs889761342 Chr.5: 179614748-179614748 on Build GRCh38
rs771208549 Chr.5: 179614817-179614817 on Build GRCh38
rs915602863 Chr.5: 179614655-179614655 on Build GRCh38
rs967367049 Chr.5: 179614781-179614781 on Build GRCh38
rs1008665544 Chr.5: 179614891-179614891 on Build GRCh38
rs114335388 Chr.5: 179614725-179614725 on Build GRCh38
rs528675391 Chr.5: 179614645-179614645 on Build GRCh38
rs554871111 Chr.5: 179614839-179614839 on Build GRCh38
rs1023660865 Chr.5: 179614591-179614594 on Build GRCh38
rs1045647388 Chr.5: 179614681-179614681 on Build GRCh38
rs954283716 Chr.5: 179614511-179614511 on Build GRCh38
rs904814271 Chr.5: 179614824-179614824 on Build GRCh38
rs142134683 Chr.5: 179614785-179614787 on Build GRCh38
rs776732707 Chr.5: 179614895-179614895 on Build GRCh38
rs750283108 Chr.5: 179614914-179614914 on Build GRCh38
rs969675817 Chr.5: 179614819-179614819 on Build GRCh38
rs562312644 Chr.5: 179614931-179614931 on Build GRCh38
rs920547242 Chr.5: 179614662-179614662 on Build GRCh38
rs999895303 Chr.5: 179614608-179614608 on Build GRCh38
rs915674977 Chr.5: 179614807-179614807 on Build GRCh38
rs572826426 Chr.5: 179614844-179614844 on Build GRCh38
rs1019791242 Chr.5: 179614765-179614765 on Build GRCh38
rs528600577 Chr.5: 179614755-179614756 on Build GRCh38
rs533719736 Chr.5: 179614860-179614860 on Build GRCh38
rs987038409 Chr.5: 179614519-179614519 on Build GRCh38

View Full Product Details