The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004708.3 | NP_004699.1 | 1 - 2 | 141 | 74 |
XM_005259392.4 | XP_005259449.1 | 1 - 2 | 168 | 74 | |
GenBank mRNA | AB209040.1 | - | 1 - 2 | 130 | 74 |
AF014955.1 | - | 1 - 2 | 91 | 74 | |
AK293486.1 | - | 1 - 2 | 139 | 74 | |
BC015519.2 | - | 1 - 2 | 107 | 74 | |
BM696336.1 | - | 1 - 2 | 131 | 74 | |
BT006694.1 | - | 1 - 2 | 67 | 74 | |
CR456934.1 | - | 1 - 2 | 67 | 74 | |
DN915905.1 | - | 1 - 2 | 141 | 74 | |
DQ208400.1 | - | 1 - 2 | 72 | 74 |