The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005107.3 | NP_005098.2 | 2 - 3 | 413 | 80 |
XM_005265635.3 | XP_005265692.1 | 2 - 3 | 362 | 80 | |
XM_006713442.3 | XP_006713505.1 | 3 - 4 | 645 | 80 | |
GenBank mRNA | AB020523.1 | - | 2 - 3 | 344 | 80 |
AK023235.1 | - | 2 - 3 | 320 | 80 |