The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128610.2 | NP_001122082.1 | 1 - 2 | 276 | 104 |
XM_017022719.1 | XP_016878208.1 | 1 - 2 | 248 | 104 | |
XM_017022722.1 | XP_016878211.1 | 1 - 2 | 261 | 104 | |
GenBank mRNA | BC026954.1 | - | 1 - 2 | 226 | 104 |
BC051345.1 | - | 1 - 2 | 175 | 104 | |
D29956.1 | - | 1 - 2 | 250 | 104 |