Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24547005-24547507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319
Entrez Gene ID
9856
Gene Name
KIAA0319
Gene Aliases
DYLX2, DYX2, NMIG
UniGene
Hs.26441
Gene Chromosome Location
Chr.6: 24540131-24646211 on Build GRCh38
Transcripts
NM_001168374.1 NM_001168375.1 NM_001168376.1 NM_001168377.1 NM_001252328.1 NM_014809.3 XM_011515025.2 XM_011515026.2 XM_017011541.1 XM_017011542.1 XM_017011543.1 XM_017011544.1 XM_017011545.1 XM_017011546.1 XM_017011547.1 XM_017011548.1 XM_017011549.1 XM_017011550.1 XM_017011551.1 XM_017011552.1 XM_017011553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373479 c.3080T>C p.M1027T Chr.6: 24547304-24547304 on Build GRCh38
7373472 c.3163T>A p.S1055T Chr.6: 24547194-24547194 on Build GRCh38
483736 c.3145G>C p.G1049R Chr.6: 24547239-24547239 on Build GRCh38
1173203 c.3178A>T p.R1060* Chr.6: 24547206-24547206 on Build GRCh38
7373471 c.3190T>A p.S1064T Chr.6: 24547194-24547194 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577878212 Chr.6: 24547423-24547423 on Build GRCh38
rs1016901264 Chr.6: 24547339-24547339 on Build GRCh38
rs746481831 Chr.6: 24547369-24547369 on Build GRCh38
rs769219851 Chr.6: 24547362-24547362 on Build GRCh38
rs1061164 Chr.6: 24547133-24547133 on Build GRCh38
rs528543316 Chr.6: 24547033-24547033 on Build GRCh38
rs192530422 Chr.6: 24547418-24547418 on Build GRCh38
rs372602681 Chr.6: 24547337-24547337 on Build GRCh38
rs369585660 Chr.6: 24547310-24547310 on Build GRCh38
rs546195987 Chr.6: 24547448-24547448 on Build GRCh38
rs752615001 Chr.6: 24547296-24547296 on Build GRCh38
rs924012051 Chr.6: 24547405-24547405 on Build GRCh38
rs187212811 Chr.6: 24547138-24547138 on Build GRCh38
rs762887635 Chr.6: 24547387-24547387 on Build GRCh38
rs1025797957 Chr.6: 24547210-24547210 on Build GRCh38
rs780765662 Chr.6: 24547345-24547345 on Build GRCh38
rs765134085 Chr.6: 24547280-24547280 on Build GRCh38
rs766002523 Chr.6: 24547245-24547245 on Build GRCh38
rs763970934 Chr.6: 24547388-24547388 on Build GRCh38
rs751504655 Chr.6: 24547116-24547116 on Build GRCh38
rs894056834 Chr.6: 24547066-24547066 on Build GRCh38
rs369788086 Chr.6: 24547389-24547389 on Build GRCh38
rs201365492 Chr.6: 24547239-24547239 on Build GRCh38
rs941326856 Chr.6: 24547356-24547356 on Build GRCh38
rs746193566 Chr.6: 24547167-24547167 on Build GRCh38
rs765009054 Chr.6: 24547381-24547381 on Build GRCh38
rs770523885 Chr.6: 24547371-24547371 on Build GRCh38
rs975254386 Chr.6: 24547358-24547358 on Build GRCh38
rs770233380 Chr.6: 24547182-24547182 on Build GRCh38
rs772865811 Chr.6: 24547368-24547368 on Build GRCh38
rs929805277 Chr.6: 24547421-24547421 on Build GRCh38
rs948278463 Chr.6: 24547454-24547454 on Build GRCh38
rs746255929 Chr.6: 24547333-24547333 on Build GRCh38
rs955779451 Chr.6: 24547146-24547146 on Build GRCh38
rs543627197 Chr.6: 24547449-24547449 on Build GRCh38
rs752807361 Chr.6: 24547124-24547124 on Build GRCh38
rs761460863 Chr.6: 24547193-24547193 on Build GRCh38
rs933613924 Chr.6: 24547228-24547228 on Build GRCh38
rs776318480 Chr.6: 24547377-24547377 on Build GRCh38
rs879040027 Chr.6: 24547207-24547207 on Build GRCh38
rs776107002 Chr.6: 24547192-24547192 on Build GRCh38
rs747223701 Chr.6: 24547158-24547158 on Build GRCh38
rs754895152 Chr.6: 24547434-24547434 on Build GRCh38
rs749808855 Chr.6: 24547361-24547361 on Build GRCh38
rs755660458 Chr.6: 24547307-24547307 on Build GRCh38
rs533758509 Chr.6: 24547297-24547297 on Build GRCh38
rs529185950 Chr.6: 24547482-24547482 on Build GRCh38
rs535150129 Chr.6: 24547304-24547304 on Build GRCh38
rs184571085 Chr.6: 24547467-24547467 on Build GRCh38
rs773066531 Chr.6: 24547216-24547216 on Build GRCh38
rs566081454 Chr.6: 24547032-24547032 on Build GRCh38
rs757441676 Chr.6: 24547331-24547331 on Build GRCh38
rs541313624 Chr.6: 24547062-24547062 on Build GRCh38
rs567594992 Chr.6: 24547299-24547299 on Build GRCh38
rs555334494 Chr.6: 24547340-24547340 on Build GRCh38
rs200733699 Chr.6: 24547202-24547202 on Build GRCh38
rs200729626 Chr.6: 24547178-24547178 on Build GRCh38
rs758447360 Chr.6: 24547136-24547136 on Build GRCh38
rs34829630 Chr.6: 24547422-24547422 on Build GRCh38
rs141315468 Chr.6: 24547161-24547161 on Build GRCh38
rs541993554 Chr.6: 24547209-24547209 on Build GRCh38
rs1024265856 Chr.6: 24547045-24547045 on Build GRCh38
rs2817245 Chr.6: 24547378-24547378 on Build GRCh38
rs980008542 Chr.6: 24547103-24547103 on Build GRCh38
rs7770041 Chr.6: 24547123-24547123 on Build GRCh38
rs781434453 Chr.6: 24547332-24547332 on Build GRCh38
rs753600089 Chr.6: 24547256-24547256 on Build GRCh38
rs759338330 Chr.6: 24547259-24547259 on Build GRCh38
rs200649619 Chr.6: 24547160-24547160 on Build GRCh38

View Full Product Details