Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24556542-24556939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319
Entrez Gene ID
9856
Gene Name
KIAA0319
Gene Aliases
DYLX2, DYX2, NMIG
UniGene
Hs.26441
Gene Chromosome Location
Chr.6: 24540131-24646211 on Build GRCh38
Transcripts
NM_001168374.1 NM_001168375.1 NM_001168376.1 NM_001168377.1 NM_001252328.1 NM_014809.3 XM_011515025.2 XM_011515026.2 XM_017011541.1 XM_017011542.1 XM_017011543.1 XM_017011544.1 XM_017011545.1 XM_017011546.1 XM_017011547.1 XM_017011548.1 XM_017011549.1 XM_017011550.1 XM_017011551.1 XM_017011552.1 XM_017011553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869863 c.2824C>T p.Q942* Chr.6: 24556640-24556640 on Build GRCh38
4540343 c.2826G>A p.Q942Q Chr.6: 24556638-24556638 on Build GRCh38
6989083 c.2801G>A p.R934H Chr.6: 24556636-24556636 on Build GRCh38
245212 c.2785C>T p.R929C Chr.6: 24556679-24556679 on Build GRCh38
5765371 c.2769C>T p.C923C Chr.6: 24556695-24556695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755687148 Chr.6: 24556631-24556631 on Build GRCh38
rs770931600 Chr.6: 24556679-24556679 on Build GRCh38
rs199538325 Chr.6: 24556694-24556694 on Build GRCh38
rs752280978 Chr.6: 24556729-24556729 on Build GRCh38
rs777917501 Chr.6: 24556663-24556663 on Build GRCh38
rs746748990 Chr.6: 24556747-24556747 on Build GRCh38
rs745899546 Chr.6: 24556686-24556686 on Build GRCh38
rs189574043 Chr.6: 24556742-24556742 on Build GRCh38
rs377219427 Chr.6: 24556615-24556615 on Build GRCh38
rs567670732 Chr.6: 24556637-24556637 on Build GRCh38
rs778391373 Chr.6: 24556641-24556641 on Build GRCh38
rs113597767 Chr.6: 24556758-24556758 on Build GRCh38
rs765324071 Chr.6: 24556570-24556570 on Build GRCh38
rs931278857 Chr.6: 24556692-24556692 on Build GRCh38
rs1034303844 Chr.6: 24556883-24556883 on Build GRCh38
rs374752663 Chr.6: 24556802-24556804 on Build GRCh38
rs904192116 Chr.6: 24556796-24556796 on Build GRCh38
rs369164359 Chr.6: 24556568-24556568 on Build GRCh38
rs769749773 Chr.6: 24556762-24556762 on Build GRCh38
rs758282824 Chr.6: 24556730-24556730 on Build GRCh38
rs770094776 Chr.6: 24556693-24556693 on Build GRCh38
rs758110457 Chr.6: 24556647-24556647 on Build GRCh38
rs757792302 Chr.6: 24556678-24556678 on Build GRCh38
rs538198671 Chr.6: 24556695-24556695 on Build GRCh38
rs970362244 Chr.6: 24556793-24556793 on Build GRCh38
rs137950263 Chr.6: 24556636-24556636 on Build GRCh38
rs553796186 Chr.6: 24556862-24556862 on Build GRCh38
rs774633095 Chr.6: 24556697-24556697 on Build GRCh38
rs754418155 Chr.6: 24556711-24556711 on Build GRCh38
rs764993564 Chr.6: 24556723-24556723 on Build GRCh38
rs1014120166 Chr.6: 24556900-24556900 on Build GRCh38
rs575158681 Chr.6: 24556753-24556753 on Build GRCh38
rs374309454 Chr.6: 24556594-24556594 on Build GRCh38
rs981783481 Chr.6: 24556800-24556802 on Build GRCh38
rs950965146 Chr.6: 24556702-24556702 on Build GRCh38
rs1041180496 Chr.6: 24556721-24556721 on Build GRCh38
rs370096303 Chr.6: 24556684-24556684 on Build GRCh38
rs895722989 Chr.6: 24556898-24556898 on Build GRCh38
rs554358650 Chr.6: 24556621-24556621 on Build GRCh38
rs890422477 Chr.6: 24556622-24556622 on Build GRCh38
rs763195822 Chr.6: 24556589-24556589 on Build GRCh38
rs143728849 Chr.6: 24556655-24556655 on Build GRCh38
rs1042780579 Chr.6: 24556795-24556795 on Build GRCh38
rs144175049 Chr.6: 24556705-24556705 on Build GRCh38
rs766462374 Chr.6: 24556683-24556683 on Build GRCh38
rs753133656 Chr.6: 24556585-24556585 on Build GRCh38
rs10946705 Chr.6: 24556708-24556708 on Build GRCh38
rs944241694 Chr.6: 24556710-24556710 on Build GRCh38
rs748018832 Chr.6: 24556642-24556642 on Build GRCh38
rs762322354 Chr.6: 24556701-24556701 on Build GRCh38
rs945802834 Chr.6: 24556788-24556788 on Build GRCh38
rs781240489 Chr.6: 24556759-24556759 on Build GRCh38
rs544191311 Chr.6: 24556761-24556761 on Build GRCh38
rs181265010 Chr.6: 24556869-24556869 on Build GRCh38
rs754592069 Chr.6: 24556657-24556658 on Build GRCh38

View Full Product Details