Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24568689-24569189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319
Entrez Gene ID
9856
Gene Name
KIAA0319
Gene Aliases
DYLX2, DYX2, NMIG
UniGene
Hs.26441
Gene Chromosome Location
Chr.6: 24540131-24646211 on Build GRCh38
Transcripts
NM_001168374.1 NM_001168375.1 NM_001168376.1 NM_001168377.1 NM_001252328.1 NM_014809.3 XM_011515025.2 XM_011515026.2 XM_017011541.1 XM_017011542.1 XM_017011543.1 XM_017011544.1 XM_017011545.1 XM_017011546.1 XM_017011547.1 XM_017011548.1 XM_017011549.1 XM_017011550.1 XM_017011551.1 XM_017011552.1 XM_017011553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338036 c.2077A>G p.S693G Chr.6: 24568817-24568817 on Build GRCh38
6338035 c.2104A>G p.S702G Chr.6: 24568817-24568817 on Build GRCh38
483737 c.2092C>A p.Q698K Chr.6: 24568829-24568829 on Build GRCh38
4805097 c.2091C>A p.D697E Chr.6: 24568830-24568830 on Build GRCh38
5686777 c.1997C>G p.P666R Chr.6: 24568924-24568924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761606146 Chr.6: 24569123-24569123 on Build GRCh38
rs945856906 Chr.6: 24569161-24569161 on Build GRCh38
rs774632962 Chr.6: 24568847-24568847 on Build GRCh38
rs138629379 Chr.6: 24568846-24568846 on Build GRCh38
rs961835307 Chr.6: 24569055-24569055 on Build GRCh38
rs972891302 Chr.6: 24568736-24568736 on Build GRCh38
rs758249116 Chr.6: 24568782-24568782 on Build GRCh38
rs752583084 Chr.6: 24568780-24568780 on Build GRCh38
rs578052918 Chr.6: 24569162-24569163 on Build GRCh38
rs979854673 Chr.6: 24568994-24568994 on Build GRCh38
rs751600062 Chr.6: 24568958-24568958 on Build GRCh38
rs201466944 Chr.6: 24568789-24568789 on Build GRCh38
rs770757146 Chr.6: 24568848-24568849 on Build GRCh38
rs374927235 Chr.6: 24568940-24568940 on Build GRCh38
rs756193762 Chr.6: 24568815-24568815 on Build GRCh38
rs540555048 Chr.6: 24568945-24568945 on Build GRCh38
rs910858513 Chr.6: 24569109-24569109 on Build GRCh38
rs137884009 Chr.6: 24568809-24568809 on Build GRCh38
rs200204818 Chr.6: 24569089-24569089 on Build GRCh38
rs767093960 Chr.6: 24569134-24569134 on Build GRCh38
rs753332323 Chr.6: 24568774-24568774 on Build GRCh38
rs745919322 Chr.6: 24568811-24568811 on Build GRCh38
rs752388832 Chr.6: 24568953-24568953 on Build GRCh38
rs764907021 Chr.6: 24568779-24568779 on Build GRCh38
rs1050861378 Chr.6: 24569074-24569074 on Build GRCh38
rs574954669 Chr.6: 24568718-24568718 on Build GRCh38
rs192776462 Chr.6: 24569158-24569158 on Build GRCh38
rs558240443 Chr.6: 24569006-24569006 on Build GRCh38
rs776038867 Chr.6: 24568932-24568932 on Build GRCh38
rs575730183 Chr.6: 24568746-24568747 on Build GRCh38
rs763966559 Chr.6: 24568883-24568883 on Build GRCh38
rs919648907 Chr.6: 24569081-24569081 on Build GRCh38
rs369361045 Chr.6: 24568973-24568973 on Build GRCh38
rs746820130 Chr.6: 24568832-24568833 on Build GRCh38
rs1025180475 Chr.6: 24569029-24569029 on Build GRCh38
rs930958923 Chr.6: 24568773-24568773 on Build GRCh38
rs777919561 Chr.6: 24568785-24568785 on Build GRCh38
rs574525065 Chr.6: 24568859-24568859 on Build GRCh38
rs990064542 Chr.6: 24569043-24569043 on Build GRCh38
rs765812369 Chr.6: 24568738-24568738 on Build GRCh38
rs776492962 Chr.6: 24568878-24568879 on Build GRCh38
rs1009695946 Chr.6: 24568886-24568886 on Build GRCh38
rs751544367 Chr.6: 24568794-24568794 on Build GRCh38
rs760245107 Chr.6: 24568737-24568737 on Build GRCh38
rs759529111 Chr.6: 24568751-24568751 on Build GRCh38
rs959622061 Chr.6: 24569004-24569004 on Build GRCh38
rs1006948098 Chr.6: 24568723-24568723 on Build GRCh38
rs780226124 Chr.6: 24568835-24568835 on Build GRCh38
rs762986199 Chr.6: 24568868-24568868 on Build GRCh38
rs371500801 Chr.6: 24568908-24568908 on Build GRCh38
rs868053374 Chr.6: 24568865-24568865 on Build GRCh38
rs376155022 Chr.6: 24569056-24569056 on Build GRCh38
rs532584121 Chr.6: 24568963-24568963 on Build GRCh38
rs138160539 Chr.6: 24568813-24568813 on Build GRCh38
rs140741790 Chr.6: 24568810-24568810 on Build GRCh38
rs201729940 Chr.6: 24568956-24568956 on Build GRCh38
rs569669768 Chr.6: 24569160-24569160 on Build GRCh38
rs751303686 Chr.6: 24568995-24568995 on Build GRCh38
rs201197457 Chr.6: 24569157-24569157 on Build GRCh38
rs1025121018 Chr.6: 24569163-24569167 on Build GRCh38
rs762623296 Chr.6: 24568954-24568954 on Build GRCh38
rs1025163473 Chr.6: 24568817-24568817 on Build GRCh38
rs41271795 Chr.6: 24568778-24568778 on Build GRCh38
rs562977917 Chr.6: 24569090-24569090 on Build GRCh38
rs942758287 Chr.6: 24569122-24569122 on Build GRCh38

View Full Product Details