Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR12D2
Entrez Gene ID
26529
Gene Name
olfactory receptor family 12 subfamily D member 2 (gene/pseudogene)
Gene Aliases
DJ994E9.8, HS6M1-20
Chromosome Location
Chr.6: 29396639 - 29397671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013936.3 NP_039224.2 1 - 1 868 62
GenBank mRNA BC069123.1 - 1 - 1 807 62
BC101742.1 - 1 - 1 868 62
BC101746.1 - 1 - 1 868 62

View Full Product Details