Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC31A
Entrez Gene ID
22872
Gene Name
SEC31 homolog A, COPII coat complex component
Gene Aliases
ABP125, ABP130, HSPC275, HSPC334, SEC31L1
Chromosome Location
Chr.4: 82818509 - 82900571 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077206.3 NP_001070674.1 12 - 13 1706 90
NM_001077207.3 NP_001070675.1 12 - 13 1706 90
NM_001077208.3 NP_001070676.1 12 - 13 1706 90
NM_001191049.2 NP_001177978.1 11 - 12 1623 90
NM_001318119.1 NP_001305048.1 12 - 13 1703 90
NM_001318120.1 NP_001305049.1 14 - 15 1920 90
GenBank mRNA AB018359.1 - 12 - 13 1562 90
AB020712.1 - 14 - 15 1887 90
AF139184.1 - 11 - 12 1509 90
AK002161.1 - 12 - 13 1579 90
AK225934.1 - 12 - 13 1566 90
AK295810.1 - 11 - 12 1521 90
BC084583.1 - 12 - 13 1545 90
BC117221.1 - 12 - 13 1574 90
BC143489.1 - 12 - 13 1571 90
BC143491.1 - 12 - 13 1574 90

View Full Product Details