The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276310.1 | NP_001263239.1 | 2 - 3 | 287 | 58 |
NM_015894.3 | NP_056978.2 | 2 - 3 | 377 | 58 | |
NR_075070.1 | 2 - 3 | 377 | 58 | ||
GenBank mRNA | AK074860.1 | - | 2 - 3 | 189 | 58 |
AK094112.1 | - | 2 - 3 | 812 | 58 | |
AK298903.1 | - | 2 - 3 | 287 | 58 | |
AK310010.1 | - | 2 - 3 | 212 | 58 | |
BC009381.2 | - | 2 - 3 | 234 | 58 | |
BC025234.1 | - | 2 - 3 | 234 | 58 | |
BC047671.1 | - | 2 - 3 | 124 | 58 | |
DA161621.1 | - | 2 - 3 | 377 | 58 | |
DA181040.1 | - | 2 - 3 | 361 | 58 |