Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42935469-42935965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3
Entrez Gene ID
10695
Gene Name
canopy FGF signaling regulator 3
Gene Aliases
CAG4A, ERDA5, PRAT4A, TNRC5
UniGene
Hs.414099
Gene Chromosome Location
Chr.6: 42928002-42939287 on Build GRCh38
Transcripts
NM_001318842.1 NM_001318845.1 NM_001318847.1 NM_001318848.1 NM_006586.4 NR_134880.1 NR_134881.1 NR_134882.1 NR_134885.1 NR_134886.1 NR_134888.1 NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318842.1 NM_001318845.1 NM_001318847.1 NM_001318848.1 NM_006586.4 NR_134880.1 NR_134881.1 NR_134882.1 NR_134885.1 NR_134886.1 NR_134888.1 NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
394817 c.371A>C p.K124T Chr.6: 42935669-42935669 on Build GRCh38
1079375 c.291C>T p.I97I Chr.6: 42935589-42935589 on Build GRCh38
1079376 c.292G>A p.E98K Chr.6: 42935590-42935590 on Build GRCh38
1079377 c.362G>A p.R121Q Chr.6: 42935660-42935660 on Build GRCh38
3874149 c.326A>C p.D109A Chr.6: 42935624-42935624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045478579 Chr.6: 42935770-42935770 on Build GRCh38
rs532376229 Chr.6: 42935589-42935589 on Build GRCh38
rs969653812 Chr.6: 42935717-42935717 on Build GRCh38
rs916407491 Chr.6: 42935716-42935719 on Build GRCh38
rs779207999 Chr.6: 42935630-42935630 on Build GRCh38
rs747974124 Chr.6: 42935584-42935584 on Build GRCh38
rs770354202 Chr.6: 42935605-42935605 on Build GRCh38
rs1039977661 Chr.6: 42935530-42935530 on Build GRCh38
rs576886223 Chr.6: 42935929-42935929 on Build GRCh38
rs925490070 Chr.6: 42935555-42935555 on Build GRCh38
rs757029420 Chr.6: 42935698-42935698 on Build GRCh38
rs372611413 Chr.6: 42935719-42935719 on Build GRCh38
rs1030689587 Chr.6: 42935676-42935676 on Build GRCh38
rs372781203 Chr.6: 42935649-42935649 on Build GRCh38
rs750467042 Chr.6: 42935556-42935556 on Build GRCh38
rs748300073 Chr.6: 42935707-42935707 on Build GRCh38
rs200837590 Chr.6: 42935611-42935611 on Build GRCh38
rs113979678 Chr.6: 42935932-42935932 on Build GRCh38
rs767720442 Chr.6: 42935650-42935650 on Build GRCh38
rs1025580532 Chr.6: 42935865-42935865 on Build GRCh38
rs980200359 Chr.6: 42935746-42935746 on Build GRCh38
rs761023974 Chr.6: 42935660-42935660 on Build GRCh38
rs535929236 Chr.6: 42935934-42935934 on Build GRCh38
rs34394157 Chr.6: 42935582-42935582 on Build GRCh38
rs951492375 Chr.6: 42935873-42935873 on Build GRCh38
rs931558387 Chr.6: 42935834-42935834 on Build GRCh38
rs775277632 Chr.6: 42935851-42935851 on Build GRCh38
rs557048971 Chr.6: 42935831-42935831 on Build GRCh38
rs912718434 Chr.6: 42935498-42935498 on Build GRCh38
rs767663551 Chr.6: 42935549-42935549 on Build GRCh38
rs986861492 Chr.6: 42935901-42935901 on Build GRCh38
rs371694689 Chr.6: 42935531-42935531 on Build GRCh38
rs781036021 Chr.6: 42935793-42935793 on Build GRCh38
rs766430868 Chr.6: 42935671-42935671 on Build GRCh38
rs758033090 Chr.6: 42935588-42935588 on Build GRCh38
rs762072969 Chr.6: 42935546-42935546 on Build GRCh38
rs942835410 Chr.6: 42935506-42935506 on Build GRCh38
rs773308781 Chr.6: 42935656-42935656 on Build GRCh38
rs373064410 Chr.6: 42935680-42935680 on Build GRCh38
rs746471637 Chr.6: 42935590-42935590 on Build GRCh38
rs777419791 Chr.6: 42935696-42935696 on Build GRCh38
rs764550592 Chr.6: 42935525-42935525 on Build GRCh38
rs572663393 Chr.6: 42935827-42935827 on Build GRCh38
rs537254459 Chr.6: 42935807-42935807 on Build GRCh38
rs1001736276 Chr.6: 42935791-42935791 on Build GRCh38
rs565824394 Chr.6: 42935619-42935619 on Build GRCh38
rs749800798 Chr.6: 42935612-42935612 on Build GRCh38
rs1048666604 Chr.6: 42935844-42935844 on Build GRCh38
rs775066972 Chr.6: 42935573-42935573 on Build GRCh38
rs116075503 Chr.6: 42935679-42935679 on Build GRCh38
rs753087895 Chr.6: 42935825-42935826 on Build GRCh38
rs568302505 Chr.6: 42935806-42935806 on Build GRCh38
rs566166789 Chr.6: 42935772-42935772 on Build GRCh38
rs369185039 Chr.6: 42935704-42935704 on Build GRCh38
rs889943368 Chr.6: 42935854-42935854 on Build GRCh38
rs2234190 Chr.6: 42935692-42935692 on Build GRCh38
rs570136285 Chr.6: 42935561-42935561 on Build GRCh38
rs962937458 Chr.6: 42935836-42935836 on Build GRCh38
rs376506809 Chr.6: 42935558-42935558 on Build GRCh38
rs368598448 Chr.6: 42935560-42935560 on Build GRCh38
rs140374695 Chr.6: 42935931-42935931 on Build GRCh38
rs774826686 Chr.6: 42935643-42935643 on Build GRCh38
rs541608352 Chr.6: 42935896-42935896 on Build GRCh38
rs778652669 Chr.6: 42935564-42935564 on Build GRCh38
rs751170164 Chr.6: 42935697-42935697 on Build GRCh38

View Full Product Details