The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201325.1 | NP_001188254.1 | 2 - 3 | 298 | 105 |
NM_001201326.1 | NP_001188255.1 | 2 - 3 | 298 | 105 | |
NM_002614.4 | NP_002605.2 | 3 - 4 | 369 | 105 | |
XM_011509616.2 | XP_011507918.1 | 1 - 2 | 214 | 105 | |
XM_011509618.2 | XP_011507920.1 | 2 - 3 | 302 | 105 | |
XM_011509619.2 | XP_011507921.2 | 1 - 2 | 214 | 105 | |
XM_017001464.1 | XP_016856953.1 | 1 - 2 | 214 | 105 | |
XM_017001465.1 | XP_016856954.1 | 1 - 2 | 214 | 105 | |
XM_017001466.1 | XP_016856955.1 | 1 - 2 | 214 | 105 | |
XM_017001467.1 | XP_016856956.1 | 2 - 3 | 405 | 105 | |
GenBank mRNA | AF012281.1 | - | 2 - 3 | 245 | 105 |
AK298269.1 | - | 2 - 3 | 239 | 105 | |
AK313100.1 | - | 2 - 3 | 250 | 105 | |
BC006496.2 | - | 2 - 3 | 309 | 105 | |
BC006518.2 | - | 2 - 3 | 309 | 105 | |
BX477678.1 | - | 3 - 4 | 368 | 105 |