Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49431611-49432156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUT
Entrez Gene ID
4594
Gene Name
methylmalonyl-CoA mutase
Gene Aliases
MCM
UniGene
Hs.485527
Gene Chromosome Location
Chr.6: 49430360-49463328 on Build GRCh38
Transcripts
NM_000255.3 XM_005249143.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874624 c.2193T>G p.A731A Chr.6: 49431788-49431788 on Build GRCh38
3353875 c.2244A>C p.Q748H Chr.6: 49431737-49431737 on Build GRCh38
3921922 c.2159A>G p.N720S Chr.6: 49431822-49431822 on Build GRCh38
3874623 c.2196C>A p.A732A Chr.6: 49431785-49431785 on Build GRCh38
4155244 c.2125-2A>C p.? Chr.6: 49431858-49431858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548852414 Chr.6: 49431655-49431655 on Build GRCh38
rs560764129 Chr.6: 49431659-49431659 on Build GRCh38
rs766839518 Chr.6: 49431725-49431725 on Build GRCh38
rs1036388920 Chr.6: 49432062-49432062 on Build GRCh38
rs1039874466 Chr.6: 49431987-49431987 on Build GRCh38
rs774736275 Chr.6: 49431731-49431731 on Build GRCh38
rs748363752 Chr.6: 49431819-49431819 on Build GRCh38
rs879253851 Chr.6: 49431784-49431787 on Build GRCh38
rs748892245 Chr.6: 49431830-49431833 on Build GRCh38
rs781466735 Chr.6: 49431818-49431818 on Build GRCh38
rs1000661730 Chr.6: 49432103-49432103 on Build GRCh38
rs372245965 Chr.6: 49431763-49431763 on Build GRCh38
rs530424880 Chr.6: 49431638-49431638 on Build GRCh38
rs879253850 Chr.6: 49431784-49431785 on Build GRCh38
rs121918252 Chr.6: 49431831-49431831 on Build GRCh38
rs369071598 Chr.6: 49431868-49431868 on Build GRCh38
rs969153423 Chr.6: 49432094-49432094 on Build GRCh38
rs758170345 Chr.6: 49431871-49431871 on Build GRCh38
rs753461919 Chr.6: 49431775-49431775 on Build GRCh38
rs912295066 Chr.6: 49431971-49431971 on Build GRCh38
rs535456138 Chr.6: 49431906-49431906 on Build GRCh38
rs946931010 Chr.6: 49432021-49432021 on Build GRCh38
rs781553746 Chr.6: 49431788-49431789 on Build GRCh38
rs199521080 Chr.6: 49431677-49431677 on Build GRCh38
rs557637525 Chr.6: 49432051-49432051 on Build GRCh38
rs922102248 Chr.6: 49432001-49432001 on Build GRCh38
rs756952588 Chr.6: 49431776-49431776 on Build GRCh38
rs554433936 Chr.6: 49432071-49432071 on Build GRCh38
rs112377715 Chr.6: 49432079-49432079 on Build GRCh38
rs770205649 Chr.6: 49431820-49431820 on Build GRCh38
rs750300062 Chr.6: 49431786-49431786 on Build GRCh38
rs149971230 Chr.6: 49432074-49432074 on Build GRCh38
rs747331003 Chr.6: 49431787-49431788 on Build GRCh38
rs943823261 Chr.6: 49431985-49431985 on Build GRCh38
rs979290163 Chr.6: 49431899-49431899 on Build GRCh38
rs956335839 Chr.6: 49431921-49431921 on Build GRCh38
rs375956047 Chr.6: 49431851-49431851 on Build GRCh38
rs983998340 Chr.6: 49431780-49431780 on Build GRCh38
rs758134896 Chr.6: 49431790-49431790 on Build GRCh38
rs749610593 Chr.6: 49431821-49431821 on Build GRCh38
rs1055505756 Chr.6: 49432075-49432075 on Build GRCh38
rs778005725 Chr.6: 49431907-49431907 on Build GRCh38
rs763629567 Chr.6: 49431772-49431772 on Build GRCh38
rs987808164 Chr.6: 49431948-49431948 on Build GRCh38
rs14341 Chr.6: 49431698-49431698 on Build GRCh38
rs188796821 Chr.6: 49431844-49431844 on Build GRCh38
rs763361965 Chr.6: 49431724-49431724 on Build GRCh38
rs1049309853 Chr.6: 49432026-49432026 on Build GRCh38
rs1043918674 Chr.6: 49432041-49432041 on Build GRCh38
rs755077681 Chr.6: 49431813-49431813 on Build GRCh38
rs766126907 Chr.6: 49431877-49431877 on Build GRCh38
rs534255888 Chr.6: 49432046-49432046 on Build GRCh38
rs762165830 Chr.6: 49431679-49431679 on Build GRCh38
rs925212279 Chr.6: 49431918-49431918 on Build GRCh38
rs201963242 Chr.6: 49431785-49431785 on Build GRCh38
rs553694025 Chr.6: 49431924-49431924 on Build GRCh38
rs750168826 Chr.6: 49431873-49431873 on Build GRCh38
rs771224892 Chr.6: 49431823-49431823 on Build GRCh38
rs960855643 Chr.6: 49431735-49431735 on Build GRCh38
rs762059871 Chr.6: 49431936-49431936 on Build GRCh38
rs758811462 Chr.6: 49431783-49431784 on Build GRCh38
rs776176938 Chr.6: 49431850-49431850 on Build GRCh38
rs760186185 Chr.6: 49431739-49431739 on Build GRCh38
rs192177415 Chr.6: 49432003-49432003 on Build GRCh38
rs764878769 Chr.6: 49431870-49431870 on Build GRCh38
rs879253852 Chr.6: 49431781-49431781 on Build GRCh38
rs1013901592 Chr.6: 49432091-49432091 on Build GRCh38
rs199933641 Chr.6: 49431694-49431694 on Build GRCh38
rs959637047 Chr.6: 49431661-49431661 on Build GRCh38
rs773699218 Chr.6: 49431883-49431889 on Build GRCh38
rs779990936 Chr.6: 49431802-49431802 on Build GRCh38
rs139265503 Chr.6: 49431890-49431890 on Build GRCh38
rs763572961 Chr.6: 49431835-49431835 on Build GRCh38
rs988137193 Chr.6: 49431957-49431957 on Build GRCh38
rs1032211241 Chr.6: 49432126-49432126 on Build GRCh38
rs868855862 Chr.6: 49431685-49431685 on Build GRCh38
rs150580087 Chr.6: 49431883-49431883 on Build GRCh38
rs914182443 Chr.6: 49432009-49432009 on Build GRCh38
rs760079537 Chr.6: 49431832-49431832 on Build GRCh38
rs773680972 Chr.6: 49431717-49431717 on Build GRCh38
rs778402274 Chr.6: 49431785-49431786 on Build GRCh38
rs746950479 Chr.6: 49431805-49431805 on Build GRCh38
rs768331144 Chr.6: 49431869-49431870 on Build GRCh38

View Full Product Details