Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83118769-83119285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY1
Entrez Gene ID
23033
Gene Name
dopey family member 1
Gene Aliases
DOP1, KIAA1117, dJ202D23.2
UniGene
Hs.520246
Gene Chromosome Location
Chr.6: 83067650-83171351 on Build GRCh38
Transcripts
NM_001199942.1 NM_015018.3 XM_011535619.2 XM_017010559.1 XM_017010560.1 XM_017010561.1 XM_017010562.1 XM_017010563.1 XM_017010564.1 XM_017010565.1 XM_017010566.1 XM_017010567.1 XM_017010568.1 XM_017010569.1 XM_017010570.1 XM_017010571.1 XM_017010572.1 XM_017010573.1 XM_017010574.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6648613 c.824A>G p.H275R Chr.6: 83118931-83118931 on Build GRCh38
6252601 c.880G>T p.G294C Chr.6: 83118987-83118987 on Build GRCh38
3394326 c.856C>T p.R286* Chr.6: 83118963-83118963 on Build GRCh38
6252602 c.880G>T p.G294* Chr.6: 83118987-83118987 on Build GRCh38
139656 c.820C>T p.L274F Chr.6: 83118927-83118927 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535959883 Chr.6: 83119124-83119124 on Build GRCh38
rs144303120 Chr.6: 83118898-83118898 on Build GRCh38
rs752106966 Chr.6: 83118921-83118921 on Build GRCh38
rs764159040 Chr.6: 83118891-83118891 on Build GRCh38
rs766252076 Chr.6: 83119031-83119031 on Build GRCh38
rs779445826 Chr.6: 83118975-83118975 on Build GRCh38
rs759575193 Chr.6: 83119037-83119037 on Build GRCh38
rs376186272 Chr.6: 83119028-83119028 on Build GRCh38
rs1054088201 Chr.6: 83119102-83119102 on Build GRCh38
rs929969161 Chr.6: 83118861-83118861 on Build GRCh38
rs563495521 Chr.6: 83119085-83119085 on Build GRCh38
rs1019937258 Chr.6: 83119047-83119047 on Build GRCh38
rs542828236 Chr.6: 83119009-83119009 on Build GRCh38
rs964239285 Chr.6: 83119168-83119168 on Build GRCh38
rs766496727 Chr.6: 83118880-83118883 on Build GRCh38
rs1029807417 Chr.6: 83119163-83119163 on Build GRCh38
rs757755405 Chr.6: 83119015-83119015 on Build GRCh38
rs768984245 Chr.6: 83119018-83119018 on Build GRCh38
rs558607678 Chr.6: 83119223-83119223 on Build GRCh38
rs759654004 Chr.6: 83118869-83118869 on Build GRCh38
rs758430895 Chr.6: 83119095-83119095 on Build GRCh38
rs992667764 Chr.6: 83119250-83119250 on Build GRCh38
rs780745646 Chr.6: 83118837-83118837 on Build GRCh38
rs755548322 Chr.6: 83119028-83119028 on Build GRCh38
rs1027179223 Chr.6: 83119174-83119174 on Build GRCh38
rs144889245 Chr.6: 83118859-83118859 on Build GRCh38
rs902815076 Chr.6: 83119054-83119054 on Build GRCh38
rs551242458 Chr.6: 83118892-83118892 on Build GRCh38
rs73749704 Chr.6: 83119200-83119200 on Build GRCh38
rs767074313 Chr.6: 83118908-83118908 on Build GRCh38
rs1004434564 Chr.6: 83119195-83119196 on Build GRCh38
rs192223559 Chr.6: 83119133-83119133 on Build GRCh38
rs777232077 Chr.6: 83118941-83118941 on Build GRCh38
rs780128569 Chr.6: 83119185-83119185 on Build GRCh38
rs749710206 Chr.6: 83118950-83118950 on Build GRCh38
rs2148010 Chr.6: 83119005-83119005 on Build GRCh38
rs141256134 Chr.6: 83119247-83119247 on Build GRCh38
rs751507783 Chr.6: 83118899-83118899 on Build GRCh38
rs899572979 Chr.6: 83119123-83119123 on Build GRCh38
rs905662395 Chr.6: 83118973-83118973 on Build GRCh38
rs776985152 Chr.6: 83119021-83119021 on Build GRCh38
rs543155175 Chr.6: 83118836-83118836 on Build GRCh38
rs775384912 Chr.6: 83118875-83118875 on Build GRCh38
rs778228661 Chr.6: 83118849-83118849 on Build GRCh38
rs561018244 Chr.6: 83119122-83119122 on Build GRCh38
rs771440985 Chr.6: 83118857-83118857 on Build GRCh38
rs1007943971 Chr.6: 83119090-83119090 on Build GRCh38
rs149504271 Chr.6: 83118895-83118895 on Build GRCh38
rs964651431 Chr.6: 83118819-83118819 on Build GRCh38
rs747174352 Chr.6: 83119197-83119197 on Build GRCh38
rs980840895 Chr.6: 83119231-83119231 on Build GRCh38
rs138779958 Chr.6: 83119208-83119208 on Build GRCh38
rs755794105 Chr.6: 83118935-83118935 on Build GRCh38
rs974763576 Chr.6: 83119172-83119172 on Build GRCh38
rs746398436 Chr.6: 83118977-83118977 on Build GRCh38
rs754581749 Chr.6: 83118900-83118900 on Build GRCh38
rs769280819 Chr.6: 83118845-83118845 on Build GRCh38
rs371305387 Chr.6: 83119033-83119033 on Build GRCh38
rs371146281 Chr.6: 83119015-83119015 on Build GRCh38
rs927159155 Chr.6: 83119232-83119232 on Build GRCh38
rs772117715 Chr.6: 83118870-83118870 on Build GRCh38
rs1009887023 Chr.6: 83119043-83119043 on Build GRCh38
rs760991326 Chr.6: 83118885-83118885 on Build GRCh38
rs949450865 Chr.6: 83118932-83118932 on Build GRCh38
rs760835052 Chr.6: 83118841-83118847 on Build GRCh38
rs550317359 Chr.6: 83119040-83119040 on Build GRCh38
rs559480659 Chr.6: 83118855-83118855 on Build GRCh38
rs77548299 Chr.6: 83118951-83118951 on Build GRCh38
rs757587957 Chr.6: 83118957-83118957 on Build GRCh38
rs113532415 Chr.6: 83118814-83118814 on Build GRCh38

View Full Product Details