Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127439323-127439866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0408
Entrez Gene ID
9729
Gene Name
KIAA0408
Gene Aliases
-
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127459390 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Gene Symbol
SOGA3
Entrez Gene ID
387104
Gene Name
SOGA family member 3
Gene Aliases
C6orf174, dJ403A15.3
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127519355 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

SNP Detail

SNP ID Chromosome Location
rs1055554608 Chr.6: 127439695-127439695 on Build GRCh38
rs561451574 Chr.6: 127439612-127439612 on Build GRCh38
rs748066391 Chr.6: 127439731-127439735 on Build GRCh38
rs938627491 Chr.6: 127439777-127439777 on Build GRCh38
rs9321091 Chr.6: 127439652-127439652 on Build GRCh38
rs764213542 Chr.6: 127439702-127439702 on Build GRCh38
rs975532287 Chr.6: 127439507-127439507 on Build GRCh38
rs967174891 Chr.6: 127439375-127439375 on Build GRCh38
rs542771949 Chr.6: 127439611-127439611 on Build GRCh38
rs950783027 Chr.6: 127439353-127439353 on Build GRCh38
rs967286162 Chr.6: 127439820-127439820 on Build GRCh38
rs1011808091 Chr.6: 127439630-127439630 on Build GRCh38
rs1046994734 Chr.6: 127439627-127439627 on Build GRCh38
rs1047770762 Chr.6: 127439812-127439812 on Build GRCh38
rs745878667 Chr.6: 127439421-127439421 on Build GRCh38
rs3938 Chr.6: 127439350-127439350 on Build GRCh38
rs78312064 Chr.6: 127439628-127439628 on Build GRCh38
rs1044732892 Chr.6: 127439701-127439701 on Build GRCh38
rs565528714 Chr.6: 127439744-127439744 on Build GRCh38
rs1011259614 Chr.6: 127439713-127439713 on Build GRCh38
rs1027976790 Chr.6: 127439473-127439473 on Build GRCh38
rs117623649 Chr.6: 127439813-127439813 on Build GRCh38
rs1006357193 Chr.6: 127439377-127439377 on Build GRCh38
rs914428240 Chr.6: 127439653-127439653 on Build GRCh38
rs796930587 Chr.6: 127439627-127439627 on Build GRCh38
rs905392410 Chr.6: 127439629-127439629 on Build GRCh38
rs986447817 Chr.6: 127439543-127439543 on Build GRCh38
rs60309279 Chr.6: 127439615-127439615 on Build GRCh38
rs185072678 Chr.6: 127439532-127439532 on Build GRCh38
rs964988183 Chr.6: 127439426-127439426 on Build GRCh38
rs948644371 Chr.6: 127439663-127439663 on Build GRCh38
rs912436995 Chr.6: 127439474-127439474 on Build GRCh38
rs907140198 Chr.6: 127439764-127439764 on Build GRCh38
rs36102761 Chr.6: 127439494-127439495 on Build GRCh38
rs879501866 Chr.6: 127439478-127439478 on Build GRCh38
rs1047278006 Chr.6: 127439590-127439590 on Build GRCh38
rs528738561 Chr.6: 127439626-127439626 on Build GRCh38
rs375266035 Chr.6: 127439625-127439626 on Build GRCh38
rs1006759075 Chr.6: 127439828-127439828 on Build GRCh38
rs931334198 Chr.6: 127439585-127439585 on Build GRCh38
rs975425227 Chr.6: 127439456-127439456 on Build GRCh38

View Full Product Details